    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KIAA1324L Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          KIAA1324L Polyclonal Antibody
          View all (2) KIAA1324L antibodies
          Tech Support
          Cite KIAA1324L Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          KIAA1324L Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          KIAA1324L Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          KIAA1324L Antibody (PA5-24451) in IHC
          Immunohistochemistry analysis in formalin-fixed, paraffin-embedded human esophagus carcinoma using a KIAA1324L polyclonal antibody (Product # PA5-24451), followed by HRP-conjugated secondary antibody and DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KIAA1324L Antibody in Immunohistochemistry (IHC)
          KIAA1324L Antibody in Western Blot (WB)
          KIAA1324L Antibody in Western Blot (WB)
          KIAA1324L Antibody in Flow Cytometry (Flow)
          Product Details
          PA5-24451
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:100
          -
          Flow Cytometry (Flow)
          1:10-1:50
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          KLH conjugated synthetic peptide between 109-138 amino acids from the N-terminal region of human KIAA1324L
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          Lot-specific
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Wet ice
          RRID
          AB_2541951
          Product Specific Information
          This antibody is predicted to react with bovine based on sequence homology.
          Target Information
          KIAA1324L is a protein coding gene.
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline