FIGURE: 1 / 4
ASMT belongs to the methyltransferase superfamily, and is located in the pseudoautosomal region (PAR) at the end of the short arms of the X and Y chromosomes. The encoded enzyme catalyzes the final reaction in the synthesis of melatonin, and is abundant in the pineal gland.
Protein Aliases: acetylserotonin methyltransferase (Y chromosome); acetylserotonin N-methyltransferase; Acetylserotonin O-methyltransferase; HIOMT; Hydroxyindole O-methyltransferase; unnamed protein product
Gene Aliases: ASMT; ASMTY; HIOMT; HIOMTY
UniProt ID: (Human) P46597
Entrez Gene ID: (Human) 438