    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GPR126 Antibodies
          Invitrogen
          GPR126 Polyclonal Antibody
          View all (12) GPR126 antibodies
          Tech Support
          Cite GPR126 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          GPR126 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GPR126 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          GPR126 Antibody (PA5-50974) in WB
          Western Blot analysis of GPR126 was performed using HuvEc cells. The membrane was probed with GPR126 Polyclonal Antibody (Product # PA5-50974). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GPR126 Antibody in Western Blot (WB)
          GPR126 Polyclonal Antibody
          Product Details
          PA5-50974
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Immunohistochemistry (IHC)
          1:100-1:300
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:1,000
          -
          ELISA (ELISA)
          1:5,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthesized peptide derived from the Internal region of human GPR126 (aa 431-480).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 0.5% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2636422
          Product Specific Information
          GPR126 Polyclonal Antibody detects endogenous levels of GPR126 protein.
          Target Information
          GPR126 encodes a product that is upregulated in human umbilical vein endothelial cells, encodes a G protein-coupled receptor. Variations in this gene can affect a persoNULLs stature. Multiple transcript variants encoding different proteins have been found for this gene.
          Bioinformatics
          Protein Aliases: Adhesion G-protein coupled receptor G6; Developmentally regulated G-protein-coupled receptor; G-protein coupled receptor 126; HBV PreS1-transactivated protein 2; PS1TP2; unnamed protein product; Vascular inducible G protein-coupled receptor; vascular-inducible G protein-coupled receptor
          View more View less
          Gene Aliases: ADGRG6; APG1; DREG; GPR126; LCCS9; PR126; PS1TP2; VIGR
          View more View less
          UniProt ID: (Human) Q86SQ4
          View more View less
          Entrez Gene ID: (Human) 57211
          View more View less
          Function(s)
          endopeptidase activity transmembrane signaling receptor activity G-protein coupled receptor activity collagen binding laminin binding metal ion binding extracellular matrix binding
          Process(es)
          placenta development signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway nervous system development Schwann cell differentiation myelination in peripheral nervous system regulation of bone mineralization myelination cartilage development heart trabecula formation establishment of blood-brain barrier regulation of sprouting angiogenesis
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline