Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Protein Aliases: homeo box C11a; homeobox gene C-11; Homeobox protein Hox-C11a; HXCBA
Gene Aliases: hoxc11; hoxc11a; id:ibd3176; zgc:110834
UniProt ID: (Zebrafish) Q58EL0
Entrez Gene ID: (Zebrafish) 30403