Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PAR2 Antibodies

          Alomone Labs, LTD

          PAR2 (F2RL1) Polyclonal Antibody

          View all (22) PAR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PAR2 (F2RL1) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          PAR2 (F2RL1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PAR2 (F2RL1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PAR2 (F2RL1) Antibody (APR-032-200UL) in ICC/IF

          Expression of PAR-2 in human prostate carcinoma cell line - Immunocytochemical staining of paraformaldehyde-fixed and permeabilized human PC-3 cells. A. Cells were stained with Anti-PAR2 (F2RL1) Antibody (#APR-032), (1:500), followed by goat- Anti-rabbit-AlexaFluor-555 secondary Antibody (orange). B. Nuclei were visualized with the cell-permeable dye Hoechst 33342 (blue). C. Merge of A and B. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PAR2 (F2RL1) Antibody in Immunocytochemistry (ICC/IF)
          PAR2 (F2RL1) Antibody in Western Blot (WB)
          PAR2 (F2RL1) Antibody in Western Blot (WB)
          PAR2 (F2RL1) Antibody in Western Blot (WB)
          PAR2 (F2RL1) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PAR2 (F2RL1) Polyclonal Antibody

          Product Details

          APR-032-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)KRMQISLTSNKFSRK, corresponding to amino acid residues 368-382 of rat PAR-2, Intracellular, C-terminus
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.6 mg/mL once reconstituted

          Amount

          120 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: coagulation factor II (thrombin) receptor-like 1; coagulation factor II receptor-like 1; G-protein coupled receptor 11; G-protein-coulped receptor; PAR-2; protease-activated receptor 2; Protease-activated receptor-2; Proteinase-activated receptor-2, G protein-coupled receptor 11; thrombin receptor-like 1; unnamed protein product

          View more View less

          Gene Aliases: Gpcr11; GPR11; PAR-2; PAR2

          View more View less

          UniProt ID: (Human) P55085, (Rat) Q63645, (Mouse) P55086

          View more View less

          Entrez Gene ID: (Human) 2150, (Rat) 116677, (Mouse) 14063

          View more View less

          Function(s)
          proteinase activated receptor activity G-protein alpha-subunit binding protease binding G-protein coupled receptor activity receptor binding protein binding thrombin receptor activity G-protein beta-subunit binding signaling receptor activity heterotrimeric G-protein binding
          Process(es)
          T cell activation involved in immune response positive regulation of leukocyte chemotaxis positive regulation of cytokine production involved in immune response regulation of glomerular filtration positive regulation of glomerular filtration G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration blood coagulation negative regulation of tumor necrosis factor-mediated signaling pathway antimicrobial humoral response regulation of blood coagulation positive regulation of cell migration positive regulation of actin filament depolymerization positive regulation of pseudopodium assembly negative regulation of chemokine production positive regulation of chemokine production positive regulation of interferon-gamma production positive regulation of interleukin-1 beta production positive regulation of interleukin-10 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of superoxide anion generation positive regulation of toll-like receptor 2 signaling pathway negative regulation of toll-like receptor 3 signaling pathway positive regulation of toll-like receptor 3 signaling pathway positive regulation of toll-like receptor 4 signaling pathway positive regulation of Rho protein signal transduction neutrophil activation vasodilation regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of eosinophil degranulation cell-cell junction maintenance positive regulation of vasoconstriction positive regulation of transcription from RNA polymerase II promoter regulation of JNK cascade negative regulation of JNK cascade positive regulation of JNK cascade leukocyte migration positive regulation of chemotaxis positive regulation of positive chemotaxis defense response to virus positive regulation of protein kinase B signaling positive regulation of phagocytosis, engulfment establishment of endothelial barrier positive regulation of ERK1 and ERK2 cascade thrombin receptor signaling pathway leukocyte proliferation mature conventional dendritic cell differentiation positive regulation of renin secretion into blood stream regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of GTPase activity positive regulation of neutrophil mediated killing of gram-negative bacterium
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-nqctq:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0