Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • PARD3 Antibodies

          Abnova

          PARD3 Polyclonal Antibody, MaxPab™

          View all (10) PARD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PARD3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          PARD3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PARD3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PARD3 Antibody (H00056288-D01P) in WB

          Western Blot analysis of PARD3 expression in transfected 293T cell line (H00056288-T01) by PARD3 MaxPab polyclonal antibody.

          Lane 1: PARD3 transfected lysate(149.70 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PARD3 Antibody in Western Blot (WB)
          PARD3 Antibody in Proximity Ligation Assay (PLA) (PLA)
          PARD3 Polyclonal Antibody, MaxPab™

          Product Details

          H00056288-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Proximity Ligation Assay (PLA) (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PARD3 (AAH71566.1, 1 a.a. approximately 1340 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MKVTVCFGRT RVVVPCGDGH MKVFSLIQQA VTRYRKAIAK DPNYWIQVHR LEHGDGGILD LDDILCDVAD DKDRLVAVFD EQDPHHGGDG TSASSTGTQS PEIFGSELGT NNVSAFQPYQ ATSEIEVTPS VLRANMPLHV RRSSDPALIG LSTSVSDSNF SSEEPSRKNP TRWSTTAGFL KQNTAGSPKT CDRKKDENYR SLPRDTSNWS NQFQRDNARS SLSASHPMVG KWLEKQEQDE DGTEEDNSRV EPVGHADTGL EHIPNFSLDD MVKLVEVPND GGPLGIHVVP FSARGGRTLG LLVKRLEKGG KAEHENLFRE NDCIVRINDG DLRNRRFEQA QHMFRQAMRT PIIWFHVVPA ANKEQYEQLS QSEKNNYYSS RFSPDSQYID NRSVNSAGLH TVQRAPRLNH PPEQIDSHSR LPHSAHPSGK PPSAPASAPQ NVFSTTVSSG YNTKKIGKRL NIQLKKGTEG LGFSITSRDV TIGGSAPIYV KNILPRGAAI QDGRLKAGDR LIEVNGVDLV GKSQEEVVSL LRSTKMEGTV SLLVFRQEDA FHPRELKAED EDIVLTPDGT REFLTFEVPL NDSGSAGLGV SVKGNRSKEN HADLGIFVKS IINGGAASKD GRLRVNDQLI AVNGESLLGK TNQDAMETLR RSMSTEGNKR GMIQLIVARR ISKCNELKSP GSPPGPELPI ETALDDRERR ISHSLYSGIE GLDESPSRNA ALSRIMGKYQ LSPTVNMPQD DTVIIEDNRL PVLPPHLSDQ SSSSSHDDVG FVTADAGTWA KAAISDSADC SLSPDVDPVL AFQREGFGRQ SMSEKRTKQF SDASQLDFVK TRKSKSMDLG IADETKLNTV DDQKAGSPSR DVGPSLGLKK SSSLESLQTA VAEVTLNGDI PFHRPRPRII RGRGCNESFR AAIDKSYDKP AVDDDDEGME TLEEDTEESS RSGRESVSTA SDQPSHSLER QMNGNQEKGD KTDRKKDKTG KEKKKDRDKE KDKMKAKKGM LKGLGDMFRF GKHRKDDKIE KTGKIKIQES FTSEEERIRM KQEQERIQAK TREFRERQAR ERDYAEIQDF HRTFGCDDEL MYGGVSSYEG SMALNARPQS PREGHMMDAL YAQVKKPRNS KPSPVDSNRS TPSNHDRIQR LRQEFQQAKQ DEDVEDRRRT YSFEQPWPNA RPATQSGRHS VSVEVQMQRQ RQEERESSQQ AQRQYSSLPR QSRKNASSVS QDSWEQNYSP GEGFQSAKEN PRYSSYQGSR NGYLGGHGFN ARVMLETQEL LRQEQRRKEQ QMKKQPPSEG PSNYDSYKKV QDPSYAPPKG PFRQDVPPSP SQVARLNRLQ TPEKGRPFYS

          Target Information

          This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ASIP; ASIPLB; ASIPSB; atypical PKC isotype-specific interacting protein; Atypical PKC isotype-specific-interacting protein; bazooka; CTCL tumor antigen se2-5; FLJ21015; hASIPla; hASIPsa; multi-PDZ containing protein; PAR3; PAR-3; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3-alpha; PAR3A; multi-PDZ domain containing protein; PAR3B; multi-PDZ domain containing protein; PAR3D; multi-PDZ domain containing protein; PAR3E; multi-PDZ domain containing protein; PAR3F; multi-PDZ domain containing protein; Partitioning defective 3 homolog; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1; unnamed protein product

          View more View less

          Gene Aliases: ASIP; Baz; PAR3; PAR3A; PAR3alpha; PARD-3; PARD3; PARD3A; PPP1R118; SE2-5L16; SE2-5LT1; SE2-5T2

          View more View less

          UniProt ID: (Human) Q8TEW0

          View more View less

          Entrez Gene ID: (Human) 56288

          View more View less

          Function(s)
          protein binding phosphatidylinositol-4,5-bisphosphate binding phosphatidylinositol-3,4,5-trisphosphate binding lipid binding phosphatidylinositol-3-phosphate binding phosphatidylinositol binding
          Process(es)
          microtubule cytoskeleton organization protein targeting to membrane cell adhesion establishment or maintenance of cell polarity phospholipase C-activating G-protein coupled receptor signaling pathway axonogenesis protein localization asymmetric cell division negative regulation of peptidyl-threonine phosphorylation myelination in peripheral nervous system establishment of cell polarity cell differentiation positive regulation of myelination establishment or maintenance of epithelial cell apical/basal polarity cell division establishment of centrosome localization protein-containing complex assembly bicellular tight junction assembly establishment of epithelial cell polarity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline