PA1-27663 detects PARG1 from human samples.
PA1-27663 may be successfully used in Western blot applications.
The immunogen is a synthetic peptide corresponding to human PTPL1-associated RhoGAP.
Rap1 is a small GTPase that, through effectors, regulates Rho GTPase signaling. These effectors- Rasip1, Radil, and the protein encoded by this gene- translocate to the cell membrane, where they form a multiprotein complex. This complex is necessary for Rap1-induced inhibition of Rho signaling. Defects in this gene may be a cause of nonsyndromic cleft lip with or without cleft palate.