Recommended positive controls: HepG2.
Predicted reactivity: Cow: 92%; Dog: 91%; Horse: 100%; Mouse: 77%; Pig: 85%; Rabbit: 100%; Rat: 92%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.