Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PAX2 Antibodies

          Invitrogen

          PAX2 Polyclonal Antibody

          70 Published Figures
          184 References
          View all (29) PAX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PAX2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (70)
          PAX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PAX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 71

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PAX2 Antibody (71-6000) in WB

          Western blot of mouse embryonic kidney using Rabbit anti-Pax-2 Polyclonal Antibody (Product # 71-6000). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PAX2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in In Situ Hybridization (ISH)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PAX2 Polyclonal Antibody

          Product Details

          71-6000

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 97 publications 97 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 21 publications 21 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 11 publications 11 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 28 publications 28 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Sheep, Rat, Xenopus, Zebrafish

          Published species

          Chicken, Cynomolgus monkey, Dog, Human, Mouse, Rat, Rhesus monkey, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A GST-Pax-2 fusion protein derived from the C-terminal domain (aa188-385) of the murine Pax-2 protein
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533990

          Target Information

          The pax genes are a family of transcription factors that are active in specific tissues during early embryonic development. Pax family members possess a DNA-binding domain encoded by the paired box. Because paired domain containing genes encode transcription factors, they are capable of executing a genetic program. Several pax genes have also been associated with developmental mutations including: pax-3, which is associated with Waardenburg syndrome, pax-6, which is associated with Aniridia, and pax-2, which is associated with Wilms tumor. During embryogenesis, Pax-2 is expressed in the developing kidney. In particular, the pax-2 gene is expressed in condensing metanephric mesenchyme and in early epithelial structures derived from mesenchyme; however, pax-2 mRNA and protein levels are rapidly down regulated as the tubular epithelium matures. Although Pax-2 is down regulated during renal epithelium maturation, Pax-2 expression persists in the undifferentiated epithelium of Wilms' tumors. Persistent expression of Pax-2 in Wilms' tumors occurs frequently and correlates with the proliferation of poorly differentiated epithelial cells in these tumors. Interestingly, expression of the Wilms' tumor suppresser protein, WT1, coincides with down-regulation of the pax-2 gene; thus, suggesting that WT1 can directly repress pax-2 transcription.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: optic disc coloboma; paired box gene 2; paired box homeotic gene 2; paired box protein 2

          View more View less

          Gene Aliases: FSGS7; Opdc; PAPRS; PAX-2

          View more View less

          UniProt ID: (Human) Q02962, (Mouse) P32114

          View more View less

          Entrez Gene ID: (Sheep) 100422802, (Human) 5076, (Mouse) 18504, (Rat) 293992

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding transcription factor binding sequence-specific double-stranded DNA binding C2H2 zinc finger domain binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter urogenital system development metanephros development ureteric bud development branching involved in ureteric bud morphogenesis cell fate determination kidney development mesonephros development neural tube closure optic cup morphogenesis involved in camera-type eye development mesenchymal to epithelial transition involved in metanephros morphogenesis retinal pigment epithelium development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter nervous system development central nervous system development sensory organ development mesodermal cell fate specification positive regulation of cell proliferation glial cell differentiation optic nerve development optic nerve morphogenesis optic nerve structural organization vestibulocochlear nerve formation pancreas development regulation of metanephros size ureter maturation pronephric field specification inner ear morphogenesis regulation of apoptotic process camera-type eye development negative regulation of apoptotic process regulation of programmed cell death negative regulation of programmed cell death negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter embryonic organ morphogenesis system development pronephros development brain morphogenesis stem cell differentiation positive regulation of epithelial cell proliferation mesenchymal to epithelial transition ureteric bud morphogenesis paramesonephric duct development optic chiasma development cellular response to mechanical stimulus cellular response to retinoic acid cellular response to glucose stimulus dopaminergic neuron differentiation metanephric mesenchyme development positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis metanephric mesenchymal cell differentiation mesonephric tubule development mesonephric tubule formation mesonephric duct development nephric duct formation ureter development ureter morphogenesis metanephric collecting duct development metanephric epithelium development metanephric distal convoluted tubule development metanephric nephron tubule formation positive regulation of metanephric glomerulus development regulation of metanephric nephron tubule epithelial cell differentiation cochlea development cochlea morphogenesis positive regulation of branching involved in ureteric bud morphogenesis negative regulation of reactive oxygen species metabolic process positive regulation of metanephric DCT cell differentiation positive regulation of optic nerve formation response to nutrient levels cellular response to epidermal growth factor stimulus axonogenesis visual perception cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-dg9d8:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0