Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PCSK5 Antibodies

          Proteintech

          PC6 Polyclonal Antibody

          2 References
          View all (4) PCSK5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PC6 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (19)
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PC6 Antibody (16470-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using (Product # 16470-1-AP) (PCSK5 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PC6 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PC6 Polyclonal Antibody

          Product Details

          16470-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PC6 Fusion Protein Ag9552 (32-351 aa encoded by BC012064)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.18 mg/mL

          Amount

          27 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TRVYTNHWA VKIAGGFPEA NRIASKYGFI NIGQIGALKD YYHFYHSRTI KRSVISSRGT HSFISMEPKV EWIQQQVVKK RTKRDYDFSR AQSTYFNDPK WPSMWYMHCS DNTHPCQSDM NIEGAWKRGY TGKNIVVTIL DDGIERTHPD LMQNYDALAS CDVNGNDLDP MPRYDASNEN KHGTRCAGEV AAAANNSHCT VGIAFNAKIG GVRMLDGDVT DMVEAKSVSF NPQHVHIYSA SWGPDDDGKT VDGPAPLTRQ AFENGVRMGR RGLGSVFVWA SGNGGRSKDH CSCDGYTNSI YTISISSTAE SGKKPWYLEE C (32-351 aa encoded by BC012064)

          Target Information

          PCSK5 belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. PCSK5 mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known. The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: prohormone convertase 5; proprotein convertase 5; proprotein convertase 6; proprotein convertase PC5; protease PC6; subtilase; subtilisin-like proprotein convertase 6; subtilisin/kexin-like protease PC5; unnamed protein product

          View more View less

          Gene Aliases: b2b1549Clo; b2b585Clo; PC5; PC6; PC6A; SPC6

          View more View less

          UniProt ID: (Human) Q92824, (Mouse) Q04592

          View more View less

          Entrez Gene ID: (Human) 5125, (Mouse) 18552

          View more View less

          Function(s)
          endopeptidase activity serine-type endopeptidase activity endopeptidase inhibitor activity protein binding peptidase activity serine-type peptidase activity peptide binding
          Process(es)
          kidney development cardiac septum development proteolysis determination of left/right symmetry heart development embryo implantation anterior/posterior pattern specification protein processing peptide hormone processing viral life cycle respiratory tube development negative regulation of low-density lipoprotein particle receptor catabolic process positive regulation of integrin activation limb morphogenesis cholesterol homeostasis peptide biosynthetic process embryonic digestive tract development embryonic skeletal system development coronary vasculature development cytokine precursor processing positive regulation of vascular associated smooth muscle cell migration positive regulation of smooth muscle cell-matrix adhesion positive regulation of integrin-mediated signaling pathway renin secretion into blood stream cell-cell signaling secretion by cell plasma lipoprotein particle remodeling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-8cpz7:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch