Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TPIIQEYFE HGDTNEVAEM LRDLNLGEMK SGVPVLAVSL ALEGKASHRE MTSKLLSDLC GTVMSTTDVE KSFDKLLKDL PELALDTPRA PQLVGQFIAR AVGDGILCNT YIDSYKGTVD CVQARAALDK ATVLLSMSKG GKRKDSVWGS GGGQQSVNHL VKEIDMLLKE YLLSGDISEA EHCLKELEVP HFHHELVYEA IIMVLESTGE STFKMILDLL KSLWKSSTIT VDQMKRGYER IYNEIPDINL DVPHSYSVLE RFVEECFQAG IISKQLRDLC PSRGRKRFVS EGDGGRLKPE SY (169-469 aa encoded by BC026104)
Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. During this process, the expression patterns of numerous genes are radically altered. One such gene is the programmed cell death protein 4 (PDCD4), whose expression was found to be upregulated in all cell lines following the onset of apoptosis. PDCD4 encodes a tumor suppressor protein whose expression is lost in carcinomas of breast, colon, lung and prostate. It can bind to and inhibit the helicase activity of the eukaryotic translation initiation factor 4A and inhibit the transactivation and transformation mediated by the transcription factor AP-1. The kinase Akt regulates PDCD4 by phosphorylation, decreasing the ability of PDCD4 to interfere with the transactivation of AP-1-responsive promoter by c-Jun. There are two known isoforms of PDCD4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: death up-regulated gene protein; death-upregulated; DUG; MA-3; MGC33046; MGC33047; neoplastic transformation inhibitor protein; nuclear antigen H731; Pdcd4; programmed cell death 4 (neoplastic transformation inhibitor); protein 197/15a; protein MA-3; RP11-348N5.4; TIS; topoisomerase-inhibitor suppressed protein; unnamed protein product
Gene Aliases: D19Ucla1; Dug; H731; Ma3; Tis
UniProt ID: (Human) Q53EL6, (Rat) Q9JID1, (Mouse) Q61823
Entrez Gene ID: (Human) 27250, (Rat) 64031, (Mouse) 18569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support