Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PDE4D3 Antibodies

          Invitrogen

          PDE4D3 Polyclonal Antibody

          View all (4) PDE4D3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PDE4D3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          PDE4D3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PDE4D3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PDE4D3 Antibody (PA1-31137) in WB

          Western Blot analysis of PDE4D3 recombinant protein using PDE4D3 Polyclonal Antibody (Product # PA1-31137). Dilution: 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PDE4D3 Antibody in Western Blot (WB)
          PDE4D3 Antibody in Western Blot (WB)

          Product Details

          PA1-31137

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunohistochemistry (IHC)

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:200
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:150
          -
          Product Specifications

          Species Reactivity

          Chicken, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to unique amino acid sequence on PDE4D3 variant.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.68 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          tris glycine, pH 7.4-7.9, with 30% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1959392

          Product Specific Information

          Recommended positive controls: Rat cortical membranes.



          PA1-31137 does not cross react with PDE4D1, D2, D4 and D5. Does not cross react with PDE4A, PDE4B or PDE4C.

          Target Information

          PDE4D3 (Phosphodiesterase 4D, cAMP-specific, isoform 3) is an enzyme encoded by the PDE4D gene, which is located on chromosome 5. PDE4D3 belongs to the PDE4 family, which plays a crucial role in hydrolyzing cyclic AMP (cAMP), thereby regulating intracellular levels of this important signaling molecule. PDE4D3 is integral in modulating cAMP signaling pathways, impacting processes such as cardiac function and inflammation. This isoform of phosphodiesterase has been found to interact within a large protein complex, including Epac1, PKA, and ERK5, which is anchored by mAKAP. Phosphorylation by ERK decreases PDE4D3 activity, leading to local cAMP accumulation and increased activation of cAMP-dependent pathways.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3',5'-cyclic-AMP phosphodiesterase 4D; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D; cAMP-specific phosphodiesterase splice variant PDE4D8; cyclic AMP specific phosphodiesterase PDE4D5A; DKFZp686M11213; DPDE3; dunce; FLJ97311; HGNC:8783; HS PDE4DN2; PDE4D3; PDE4D7; Phosphodiesterase 4D cAMP-specific (dunce; phosphodiesterase 4D, cAMP specific; phosphodiesterase 4D, cAMP-specific; Phosphodiesterase 4D, cAMP-specific (dunce; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila)

          View more View less

          Gene Aliases: 9630011N22Rik; Dpde3; Pde4d

          View more View less

          UniProt ID: (Rat) P14270, (Mouse) Q01063

          View more View less

          Entrez Gene ID: (Chicken) 769469, (Rat) 24627, (Mouse) 238871

          View more View less

          Function(s)
          cyclic-nucleotide phosphodiesterase activity 3',5'-cyclic-nucleotide phosphodiesterase activity 3',5'-cyclic-AMP phosphodiesterase activity protein binding drug binding SH3 domain binding enzyme binding protein domain specific binding cAMP binding ubiquitin protein ligase binding beta-2 adrenergic receptor binding ion channel binding metal ion binding ATPase binding scaffold protein binding phosphoric diester hydrolase activity hydrolase activity phosphodiesterase
          Process(es)
          regulation of heart rate cAMP catabolic process smooth muscle contraction aging memory regulation of G-protein coupled receptor protein signaling pathway regulation of receptor activity regulation of protein kinase A signaling regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum positive regulation of smooth muscle cell migration lung development neutrophil chemotaxis positive regulation of interferon-gamma production positive regulation of interleukin-2 production positive regulation of interleukin-5 production negative regulation of peptidyl-serine phosphorylation multicellular organism growth cellular protein complex assembly negative regulation of heart contraction positive regulation of smooth muscle cell proliferation T cell receptor signaling pathway leukocyte migration regulation of ryanodine-sensitive calcium-release channel activity establishment of endothelial barrier cellular response to lipopolysaccharide cellular response to cAMP cellular response to epinephrine stimulus adrenergic receptor signaling pathway regulation of cardiac muscle cell contraction adrenergic receptor signaling pathway involved in positive regulation of heart rate negative regulation of relaxation of cardiac muscle signal transduction regulation of cell communication by electrical coupling involved in cardiac conduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline