Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: LVITPPGPEF VLNISSTFVL TCSGSAPVMW EQMSQVPWQE AAMNQDGTFS SVLTLTNVTG GDTGEYFCVY NNSLGPELSE RKRIYIFVPD PTMGFLPMDS EDLFIFVTDV TETTIPCRVT DPQLEVTLHE KKVDIPLHVP YDHQRGFTGT FEDKTYICKT TIGDREVDSD TYYVYSLQVS SINVSVNAVQ TVVRQGESIT IRCIVMGNDV VNFQWTYPRM KSGRLVEPVT DYLFGVPSRI GSILHIPTAE LSDSGTYTCN VSVSVNDHGD EKAINISVIE NGYVRLLETL GDVEIAELHR SRTLRVVFEA YPMPSVLWLK DNRTLGDSGA GELVLSTRNM SETRYVSELI LVRVKVSEAG YYTMRAFHED DEVQLSFKLQ VNVPVRVLEL SESHPANGEQ TIRCRGRGMP QPNVTWSTCR DLKRCPRKLS PTPLGNSSKE ESQLETNVTF WEEDQEYEVV STLRLRHVDQ PLSVRCMLQN SMGGDSQEVT VVPHSLPFKV
PDGFRB is a receptor tyrosine kinase and has been implicated in atherogenesis and in cell transformation by the SIS oncogene. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. The gene is flanked on chromosome 5 by the genes for granulocyte-macrophage colony-stimulating factor and macrophage-colony stimulating factor receptor; all three genes may be implicated in the 5-q syndrome. A translocation between chromosomes 5 and 12, that fuses PDGFRB (CD140b) to that of the translocation, ETV6, leukemia gene, results in chronic myeloproliferative disorder with eosinophilia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140b; CD140b antigen; PDGF beta chain; PDGF Receptor beta; PDGF-R-beta; PDGFR-1; PDGFR-beta; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta
Gene Aliases: AI528809; CD140b; Pdgfr; PDGFR-1; Pdgfr1; Pdgfrb
UniProt ID: (Rat) Q05030, (Mouse) P05622
Entrez Gene ID: (Rat) 24629, (Mouse) 18596
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support