Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: CDLHRLEEG PPVTTVLTRE DGLKYYRMMQ TVRRMELKAD QLYKQKIIRG FCHLCDGQEA CCVGLEAGIN PTDHLITAYR AHGFTFTRGL SVREILAELT GRKGGCAKGK GGSMHMYAKN FYGGNGIVGA QVPLGAGIAL ACKYNGKDEV CLTLYGDGAA NQGQIFEAYN MAALWKLPCI FICENNRYGM GTSVERAAAS TDYYKRGDFI PGLRVDGMDI LCVREATRFA AAYCRSGKGP ILMELQTYRY HGHSMSDPGV SYRTREEIQE VRSKSDPIML LKDRMVNSNL ASVEELKEID VEVRKEIEDA AQFATADPEP PLEELGYHIY SSDPPFEVRG ANQWIKFKSV S (41-390 aa encoded by BC002406)
The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex. Mutations in this gene are associated with pyruvate dehydrogenase E1-alpha deficiency and X-linked Leigh syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dehydrogenase; PDHE1-A type I; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase alpha 1; pyruvate dehydrogenase alpha subunit precursor (EC 1.2.4.1); pyruvate dehydrogenase complex; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1 subunit; pyruvate dehydrogenase E1-alpha precursor; pyruvate dehydrogenase E1alpha subunit; unnamed protein product
Gene Aliases: E1alpha; PDHA; Pdha-1; PDHAD; PDHCE1A; PHE1A
UniProt ID: (Human) P08559, (Mouse) P35486
Entrez Gene ID: (Human) 5160, (Mouse) 18597, (Rat) 29554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support