Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MRWVWALLKN ASLAGAPKYI EHFSKFSPSP LSMKQFLDFG SSNACEKASF TFLRQELPVR LANIMKEINL LPDRVLSTPS VQLVQSWYVQ SLLDIMKFLD KDPEDHRTLS QFTDALVTIR NRHNDVVPTM AQGVLEYKDT YGDDPVSNQN IQYFLDRFYL SRISIRMLIN QHTLIFDGST NPAHPKHIGS IDPNCNVSEV VKDAYDMAKL LCDKYYMASP DLEIQEINAA NSKQPIHMVY VPSHLYHMLF ELFKNAMRAT VESHESSLIL PPIKVMVALG EEDLSIKMSD RGGGVPLRKI ERLFSYMYST APTPQPGTGG TPLAGFGYGL PISRLYAKYF QGDLQLFSME GFGTDAVIYL KALSTDSVER LPVYNKSAWR HYQTIQEAGD WCVPSTEPKN TSTYRVS
PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial; PDH kinase 2; pyruvate dehydrogenase; Pyruvate dehydrogenase kinase isoform 2; pyruvate dehydrogenase kinase, isoenzyme 2; pyruvate dehydrogenase, lipoamide, kinase isozyme 2, mitochondrial
Gene Aliases: PDHK2; PDK2; PDKII
UniProt ID: (Human) Q15119
Entrez Gene ID: (Human) 5164
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support