Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQPRWAQGAT MWLGVLLTLL LCSSLEGQEN SFTINSVDMK SLPDWTVQNG KNLTLQCFAD VSTTSHVKPQ HQMLFYKDDV LFYNISSMKS TESYFIPEVR IYDSGTYKCT VIVNNKEKTT AEYQVLVEGV PSPRVTLDKK EAIQGGIVRV NCSVPEEKAP IHFTIEKLEL NEKMVKLKRE KNSRDQNFVI LEFPVEEQDR VLSFRCQARI ISGIHMQTSE STKSELVTVT ESFSTPKFHI SPTGMIMEGA QLHIKCTIQV THLAQEFPEI IIQKDKAIVA HNRHGNKAVY SVMAMVEHSG NYTCKVESSR ISKVSSIVVN ITELFSKPEL ESSFTHLDQG ERLNLSCSIP GAPPANFTIQ KEDTIVSQTQ DFTKIASKSD SGTYICTAGI DKVVKKSNTV QIVVCEMLSQ PRISYDAQFE VIKGQTIEVR CESISGTLPI SYQLLKTSKV LENSTKNSND PAVFKDNPTE DVEYQCVADN CHSHAKMLSE VLRVKVIAPV DEVQISILSS KVVESGEDIV LQCAVNEGSG PITYKFYREK EGKPFYQMTS NATQAFWTKQ KANKEQEGEY YCTAFNRANH ASSVPRSKIL TVRVILAPWK KGLIAVVIIG VIIALLIIAA KCYFLRKAKA KQMPVEMSRP AVPLLNSNNE KMSDPNMEAN SHYGHNDDVG NHAMKPINDN KEPLNSDVQY TEVQVSSAES HKDLGKKDTE TVYSEVRKAV PDAVESRYSR TEGSLDGT
CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adhesion molecule; CD31; CD31 antigen; EndoCAM; GPIIA'; PECA1; PECAM-1; PECAM-1 precursor; Platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; unnamed protein product
Gene Aliases: CD31; CD31/EndoCAM; endoCAM; GPIIA'; PECA1; PECAM-1; PECAM1
UniProt ID: (Human) P16284
Entrez Gene ID: (Human) 5175
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support