Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PEG10 Antibodies

          Proteintech

          PEG10 Polyclonal Antibody

          4 References
          View all (12) PEG10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PEG10 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (19)
          PEG10 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PEG10 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PEG10 Antibody (14412-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using PEG10 antibody (Product # 14412-1-AP) at dilution of 1:0 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PEG10 Antibody in Immunocytochemistry (ICC/IF)
          PEG10 Antibody in Immunocytochemistry (ICC/IF)
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG10 Antibody in Western Blot (WB)
          PEG10 Antibody in Western Blot (WB)
          PEG10 Antibody in Western Blot (WB)
          PEG10 Polyclonal Antibody

          Product Details

          14412-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PEG10 Fusion Protein Ag5754 (1-325 aa encoded by BC050659)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.45 mg/mL

          Amount

          67.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MTERRRDEL SEEINNLREK VMKQSEENNN LQSQVQKLTE ENTTLREQVE PTPEDEDDDI ELRGAAAAAA PPPPIEEECP EDLPEKFDGN PDMLAPFMAQ CQIFMEKSTR DFSVDRVRVC FVTSMMTGRA ARWASAKLER SHYLMHNYPA FMMEMKHVFE DPQRREVAKR KIRRLRQGMG SVIDYSNAFQ MIAQDLDWNE PALIDQYHEG LSDHIQEELS HLEVAKSLSA LIGQCIHIER RLARAAAARK PRSPPRALVL PHIASHHQVD PTEPVGGARM RLTQEEKERR RKLNLCLYCG TGGHYADNCP AKASKSSPAG NSPAPL (1-325 aa encoded by B C050659)

          Target Information

          This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 2-Mar; embryonal carcinoma differentiation regulated; Embryonal carcinoma differentiation regulated protein; Embryonal carcinoma differentiation-regulated protein; mammalian retrotransposon-derived 2; Mammalian retrotransposon-derived protein 2; MEF3 like 1; MEF3-like protein 1; MmPEG10; MyEF-3; MyEF-3 like; Myelin expression factor 3; Myelin expression factor 3-like protein 1; Paternally expressed gene 10 protein; Retrotransposon gag domain-containing protein 3; retrotransposon Gag like 2; Retrotransposon-derived gag-like polyprotein; Retrotransposon-derived protein PEG10; Sushi-Ichi retrotransposon homolog 1; Ty3/Gypsy-like protein; unnamed protein product

          View more View less

          Gene Aliases: AA407948; EDR; HB-1; KIAA1051e; MAR2; MART2; MEF3L; MEF3L1; MyEF-3; PEG10; RGAG3; RTL2; SIRH1

          View more View less

          UniProt ID: (Human) Q86TG7, (Mouse) Q7TN75

          View more View less

          Entrez Gene ID: (Human) 23089, (Mouse) 170676

          View more View less

          Function(s)
          nucleic acid binding DNA binding RNA binding mRNA binding protein binding zinc ion binding metal ion binding poly(A) RNA binding
          Process(es)
          apoptotic process cell differentiation negative regulation of transforming growth factor beta receptor signaling pathway mRNA transport protein homooligomerization viral translational frameshifting vesicle-mediated intercellular transport placenta development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline