Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PERK Antibodies

          Proteintech

          PERK/EIF2AK3 Polyclonal Antibody

          68 References
          View all (43) PERK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PERK/EIF2AK3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          PERK/EIF2AK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PERK/EIF2AK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PERK/EIF2AK3 Antibody (24390-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 24390-1-AP (PERK antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PERK/EIF2AK3 Antibody in Immunocytochemistry (ICC/IF)
          PERK/EIF2AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PERK/EIF2AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PERK/EIF2AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PERK/EIF2AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PERK/EIF2AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PERK/EIF2AK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PERK/EIF2AK3 Antibody in Western Blot (WB)
          PERK/EIF2AK3 Antibody in Western Blot (WB)
          PERK/EIF2AK3 Polyclonal Antibody

          Product Details

          24390-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 63 publications 63 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PERK/EIF2AK3 Fusion Protein Ag18254 (767-1116 aa encoded by BC126354)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Amount

          49.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TDCDVEDGT MDGNDEGHSF ELCPSEASPY VRSRERTSSS IVFEDSGCDN ASSKEEPKTN RLHIGNHCAN KLTAFKPTSS KSSSEATLSI SPPRPTTLSL DLTKNTTEKL QPSSPKVYLY IQMQLCRKEN LKDWMNGRCT IEERERSVCL HIFLQIAEAV EFLHSKGLMH RDLKPSNIFF TMDDVVKVGD FGLVTAMDQD EEEQTVLTPM PAYARHTGQV GTKLYMSPEQ IHGNSYSHKV DIFSLGLILF ELLYPFSTQM ERVRTLTDVR NLKFPPLFTQ KYPCEYVMVQ DMLSPSPMER PEAINIIENA VFEDLDFPGK TVLRQRSRSL SSSGTKHSRQ SNNSHSPLPS N (767-1116 aa encoded by BC126354)

          Target Information

          EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 2.7.11.1; eIF-2 protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; pancreatic eIF-2 alpha kinase; pancreatic eIF-2alpha kinase; Pancreatic eIF2-alpha kinase; PKR-like ER kinase; PRKR-like endoplasmic reticulum kinase; Protein tyrosine kinase EIF2AK3; unnamed protein product

          View more View less

          Gene Aliases: EIF2AK3; PEK; PERK; WRS

          View more View less

          UniProt ID: (Human) Q9NZJ5

          View more View less

          Entrez Gene ID: (Human) 9451

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity eukaryotic translation initiation factor 2alpha kinase activity protein tyrosine kinase activity protein binding ATP binding kinase activity transferase activity enzyme binding protein phosphatase binding identical protein binding translation regulator activity misfolded protein binding Hsp90 protein binding protein serine kinase activity
          Process(es)
          skeletal system development ossification angiogenesis chondrocyte development translational initiation regulation of translation regulation of translational initiation response to oxidative stress ER overload response response to unfolded protein endoplasmic reticulum organization positive regulation of vascular endothelial growth factor production positive regulation of gene expression negative regulation of translation calcium-mediated signaling bone mineralization endoplasmic reticulum unfolded protein response endocrine pancreas development negative regulation of myelination negative regulation of translation in response to stress negative regulation of translational initiation in response to stress cellular response to amino acid starvation response to endoplasmic reticulum stress regulation of translation initiation in response to endoplasmic reticulum stress PERK-mediated unfolded protein response cellular response to glucose starvation proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of transcription from RNA polymerase I promoter negative regulation of translational initiation insulin-like growth factor receptor signaling pathway cellular response to cold mitochondrial respiratory chain supercomplex assembly positive regulation of protein localization to nucleus regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway response to manganese-induced endoplasmic reticulum stress
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0