Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • PEX14 Antibodies

          Proteintech

          PEX14 Polyclonal Antibody

          59 References
          View all (14) PEX14 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PEX14 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PEX14 Antibody (10594-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using PEX14 antibody (Product # 10594-1-AP) at dilution of 1:1,000 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L), (CL594-Phalloidin, red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          PEX14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEX14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEX14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEX14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEX14 Antibody in Western Blot (WB)
          PEX14 Antibody in Flow Cytometry (Flow)
          PEX14 Antibody in Immunoprecipitation (IP)
          PEX14 Polyclonal Antibody

          Product Details

          10594-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:10,000
          View 15 publications 15 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 35 publications 35 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Not applicable (N/A)

          -
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Published species

          Human, Monkey, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PEX14 Fusion Protein Ag0932 (77-377 aa encoded by BC006327)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Amount

          49.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody can be used to detect endogenous PEX14 with an apparent molecular weight of 57 kDa and recognize peroxisomal structures in human, monkey and mouse cells.

          Immunogen sequence: PSSLGPATQ VVPVQPPHLI SQPYSPAGSR WRDYGALAII MAGIAFGFHQ LYKKYLLPLI LGGREDRKQL ERMEAGLSEL SGSVAQTVTQ LQTTLASVQE LLIQQQQKIQ ELAHELAAAK ATTSTNWILE SQNINELKSE INSLKGLLLN RRQFPPSPSA PKIPSWQIPV KSPSPSSPAA VNHHSSSDIS PVSNESTSSS PGKEGHSPEG STVTYHLLGP QEEGEGVVDV KGQVRMEVQG EEEKREDKED EEDEEDDDVS HVDEEDCLGV QREDRRGGDG QINEQVEKLR RPEGASNESE RD (77-377 aa encoded by BC006327)

          Target Information

          PEX14 belongs to the peroxin 14 family. It is a component of the peroxisomal translocation machinery with PEX13 and PEX17 and interacts with both the PTS1 and PTS2 receptors. PEX14 binds directly to PEX17. Defects in PEX14 are the cause of peroxisome biogenesis disorder complementation group K (PBD-CGK) and Zellweger syndrome (ZWS).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: anchor protein for HsPex5p; dJ734G22.2; NAPP2; NF-E2 associated polypeptide 2; PBD13A; Peroxin 14; Peroxin-14; Peroxisomal membrane anchor protein PEX14; peroxisomal membrane anchor protein Pex14p; Peroxisomal membrane protein PEX14; peroxisomal protein Pex14; PEX14; Pex14p; PEXE; PTS1 receptor docking protein; PTS1 receptor-docking protein; unnamed protein product

          View more View less

          Gene Aliases: dJ734G22.2; NAPP2; PBD13A; PEX14; Pex14p; R75137

          View more View less

          UniProt ID: (Human) O75381, (Mouse) Q9R0A0, (Rat) Q642G4

          View more View less

          Entrez Gene ID: (Human) 5195, (Mouse) 56273, (Rat) 64460

          View more View less

          Function(s)
          transcription corepressor activity receptor binding protein binding microtubule binding protein transmembrane transporter activity protein binding, bridging identical protein binding beta-tubulin binding protein N-terminus binding primary active transporter
          Process(es)
          protein monoubiquitination peroxisome organization protein transport protein import into peroxisome matrix protein import into peroxisome matrix, docking protein import into peroxisome matrix, translocation protein import into peroxisome matrix, receptor recycling negative regulation of protein binding microtubule anchoring cellular response to reactive oxygen species peroxisome transport along microtubule protein unfolding negative regulation of sequence-specific DNA binding transcription factor activity protein import into peroxisome matrix, substrate release negative regulation of transcription, DNA-templated protein-containing complex assembly protein complex assembly transport protein homooligomerization negative regulation of protein homotetramerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline