Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PEX14 Antibodies

          Proteintech

          PEX14 Polyclonal Antibody, CoraLite® Plus 647

          View all (15) PEX14 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PEX14 Polyclonal Antibody, CoraLite® Plus 647

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PEX14 Antibody (CL647-10594) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite® Plus 647 PEX14 antibody (CL647-10594) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PEX14 Antibody in Immunocytochemistry (ICC/IF)
          PEX14 Antibody in Flow Cytometry (Flow)
          PEX14 Polyclonal Antibody, CoraLite® Plus 647

          Product Details

          CL647-10594

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cell suspension
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein
          3D Epitope / Immunogen

          Conjugate

          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647

          Excitation/Emission Max

          654/674 nm

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: PSSLGPATQV VPVQPPHLIS QPYSPAGSRW RDYGALAIIM AGIAFGFHQL YKKYLLPLIL GGREDRKQLE RMEAGLSELS GSVAQTVTQL QTTLASVQEL LIQQQQKIQE LAHELAAAKA TTSTNWILES QNINELKSEI NSLKGLLLNR RQFPPSPSAP KIPSWQIPVK SPSPSSPAAV NHHSSSDISP VSNESTSSSP GKEGHSPEGS TVTYHLLGPQ EEGEGVVDVK GQVRMEVQGE EEKREDKEDE EDEEDDDVSH VDEEDCLGVQ REDRRGGDGQ INEQVEKLRR PEGASNESER D

          Target Information

          PEX14 belongs to the peroxin 14 family. It is a component of the peroxisomal translocation machinery with PEX13 and PEX17 and interacts with both the PTS1 and PTS2 receptors. PEX14 binds directly to PEX17. Defects in PEX14 are the cause of peroxisome biogenesis disorder complementation group K (PBD-CGK) and Zellweger syndrome (ZWS).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: anchor protein for HsPex5p; dJ734G22.2; NAPP2; NF-E2 associated polypeptide 2; PBD13A; Peroxin 14; Peroxin-14; Peroxisomal membrane anchor protein PEX14; peroxisomal membrane anchor protein Pex14p; Peroxisomal membrane protein PEX14; peroxisomal protein Pex14; PEX14; Pex14p; PEXE; PTS1 receptor docking protein; PTS1 receptor-docking protein; unnamed protein product

          View more View less

          Gene Aliases: dJ734G22.2; NAPP2; PBD13A; PEX14; Pex14p; R75137

          View more View less

          UniProt ID: (Human) O75381, (Mouse) Q9R0A0, (Rat) Q642G4

          View more View less

          Entrez Gene ID: (Human) 5195, (Mouse) 56273, (Rat) 64460

          View more View less

          Function(s)
          transcription corepressor activity receptor binding protein binding microtubule binding protein transmembrane transporter activity protein binding, bridging identical protein binding beta-tubulin binding protein N-terminus binding primary active transporter
          Process(es)
          protein monoubiquitination peroxisome organization protein transport protein import into peroxisome matrix protein import into peroxisome matrix, docking protein import into peroxisome matrix, translocation protein import into peroxisome matrix, receptor recycling negative regulation of protein binding microtubule anchoring cellular response to reactive oxygen species peroxisome transport along microtubule protein unfolding negative regulation of sequence-specific DNA binding transcription factor activity protein import into peroxisome matrix, substrate release negative regulation of transcription, DNA-templated protein-containing complex assembly protein complex assembly transport protein homooligomerization negative regulation of protein homotetramerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline