Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • PHC1 Antibodies

          Abnova

          PHC1 Polyclonal Antibody, MaxPab™

          View all (8) PHC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PHC1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          PHC1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PHC1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PHC1 Antibody (H00001911-D01P) in WB

          Western Blot analysis of PHC1 expression in transfected 293T cell line by PHC1 MaxPab rabbit polyclonal antibody.

          Lane 1: PHC1 transfected lysate(100.20 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PHC1 Antibody in Western Blot (WB)
          PHC1 Antibody in Western Blot (WB)
          PHC1 Polyclonal Antibody, MaxPab™

          Product Details

          H00001911-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PHC1 (AAH73964.1, 1 a.a. approximately 957 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: METESEQNSN STNGSSSSGG SSRPQIAQMS LYERQAVQAT IAASRQASSP NTSTTQQQTT TTQASINLAT TSAAQLISRS QSVSSPSATT LTQSVLLGNT TSPPLNQSQA QMYLRVNRTL GRNVPLASQL ILMPNGAVAA VQQEVPSAQS PGVHADADQV QNLAVRNQQA SAQGPQMQGS TQKAIPPGAS PVSSLSQASS QALAVAQASS GATNQSLNLS QAGGGSGNSI PGSMGPGGGG QAHGGLGQLP SSGMGGGSCP RKGTGVVQPL PAAQTVTVSQ GSQTEAESAA AKKAEADGSG QQNVGMNLTR TATPAPSQTL ISSATYTQIQ PHSLIQQQQQ IHLQQKQVVI QQQIAIHHQQ QFQHRQSQLL HTATHLQLAQ QQQQQQQQQQ QQPQATTLTA PQPPQVPPTQ QVPPSQSQQQ AQTLVVQPML QSSPLSLPPD AAPKPPIPIQ SKPPVAPIKP PQLGAAKMSA AQQPPPHIPV QVVGTRQPGT AQAQALGLAQ LAAAVPTSRG MPGTVQSGQA HLASSPPSSQ APGALQECPP TLAPGMTLAP VQGTAHVVKG GATTSSPVVA QVPAAFYMQS VHLPGKPQTL AVKRKADSEE ERDDVSTLGS MLPAKASPVA ESPKVMDEKS SLGEKAESVA NVNANTPSSE LVALTPAPSV PPPTLAMVSR QMGDSKPPQA IVKPQILTHI IEGFVIQEGA EPFPVGCSQL LKESEKPLQT GLPTGLTENQ SGGSLGVDSP SAELDKKANL LKCEYCGKYA PAEQFRGSKR FCSMTCAKRY NVSCSHQFRL KRKKMKEFQE ANYARVRRRG PRRSSSDIAR AKIQGKCHRG QEDSSRGSDN SSYDEALSPT SPGPLSVRAG HGERDLGNPN TAPPTPELHG INPVFLSSNP SRWSVEEVYE FIASLQGCQE IAEEFRSQEI DGQALLLLKE EHLMSAMNIK LGPALKICAK INVLKET

          Target Information

          Polycomb group (PcG) proteins assemble into multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes throughout development. PHC1 (polyhomeotic homolog 1), also known as EDR1, HPH1 or RAE28, is a 1,004 amino acid nuclear protein that is a component of the PcG multiprotein PRC1 complex. Specifically, the PcG PRC1 complex modifies histones, remodels chromatin and mediates monoubiquination of Histone H2A. Other constituent proteins involved in the PcG PRC1 complex are Mel-18, Bmi-1, M33, MPc2, MPc3, RING1, Ring1b, as well as several others. Existing as a homodimer, PHC1 contains one FCS-type zinc finger and a SAM (sterile alpha motif) domain. PHC1 is encoded by a gene located on human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp686A1782; early development regulator 1 (homolog of polyhomeotic 1); Early development regulatory protein 1; hPH1; polyhomeotic-like 1; Polyhomeotic-like protein 1; unnamed protein product

          View more View less

          Gene Aliases: EDR1; HPH1; MCPH11; PH1; PHC1; RAE28

          View more View less

          UniProt ID: (Human) P78364

          View more View less

          Entrez Gene ID: (Human) 1911

          View more View less

          Function(s)
          DNA binding chromatin binding protein binding zinc ion binding histone binding metal ion binding
          Process(es)
          chromatin remodeling negative regulation of transcription, DNA-templated cellular response to retinoic acid cellular response to leukemia inhibitory factor
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline