Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PHEX Antibodies

          Abnova

          PHEX Polyclonal Antibody, MaxPab™

          View all (5) PHEX antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PHEX Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PHEX Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PHEX Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PHEX Antibody (H00005251-D01P) in WB

          Western Blot analysis of PHEX expression in transfected 293T cell line by PHEX MaxPab polyclonal antibody. Lane 1: PHEX transfected lysate(86.50 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PHEX Antibody in Western Blot (WB)
          PHEX Polyclonal Antibody, MaxPab™

          Product Details

          H00005251-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PHEX (NP_000435.3, 1 a.a. approximately 749 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEAETGSSVE TGKKANRGTR IALVVFVGGT LVLGTILFLV SQGLLSLQAK QEYCLKPECI EAAAAILSKV NLSVDPCDNF FRFACDGWIS NNPIPEDMPS YGVYPWLRHN VDLKLKELLE KSISRRRDTE AIQKAKILYS SCMNEKAIEK ADAKPLLHIL RHSPFRWPVL ESNIGPEGVW SERKFSLLQT LATFRGQYSN SVFIRLYVSP DDKASNEHIL KLDQATLSLA VREDYLDNST EAKSYRDALY KFMVDTAVLL GANSSRAEHD MKSVLRLEIK IAEIMIPHEN RTSEAMYNKM NISELSAMIP QFDWLGYIKK VIDTRLYPHL KDISPSENVV VRVPQYFKDL FRILGSERKK TIANYLVWRM VYSRIPNLSR RFQYRWLEFS RVIQGTTTLL PQWDKCVNFI ESALPYVVGK MFVDVYFQED KKEMMEELVE GVRWAFIDML EKENEWMDAG TKRKAKEKAR AVLAKVGYPE FIMNDTHVNE DLKAIKFSEA DYFGNVLQTR KYLAQSDFFW LRKAVPKTEW FTNPTTVNAF YSASTNQIRF PAGELQKPFF WGTEYPRSLS YGAIGVIVGH EFTHGFDNNG RKYDKNGNLD PWWSTESEEK FKEKTKCMIN QYSNYYWKKA GLNVKGKRTL GENIADNGGL REAFRAYRKW INDRRQGLEE PLLPGITFTN NQLFFLSYAH VRCNSYRPEA AREQVQIGAH SPPQFRVNGA ISNFEEFQKA FNCPPNSTMN RGMDSCRLW

          Target Information

          Peptidase that cleaves SIBLING (small integrin-binding ligand, N-linked glycoprotein)-derived ASARM peptides, thus regulating their biological activity. Cleaves ASARM peptides between Ser and Glu or Asp residues. Regulates osteogenic cell differentiation and bone mineralization through the cleavage of the MEPE-derived ASARM peptide. Promotes dentin mineralization and renal phosphate reabsorption by cleaving DMP1- and MEPE-derived ASARM peptides. Inhibits the cleavage of MEPE by CTSB/cathepsin B thus preventing MEPE degradation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HYP; Metalloendopeptidase homolog PEX; PEX; PHEX peptidase; phosphate regulating endopeptidase homolog X-linked; phosphate regulating gene with homologies to endopeptidases on the X chromosome (hypophosphatemia, vitamin D resistant rickets); Phosphate-regulating neutral endopeptidase PHEX; similar to metalloendopeptidase; unnamed protein product; Vitamin D-resistant hypophosphatemic rickets protein; X-linked hypophosphatemia protein; X-linked hypophosphatemic rickets candidate gene product

          View more View less

          Gene Aliases: HPDR; HPDR1; HYP; HYP1; LXHR; PEX; PHEX; XLH

          View more View less

          UniProt ID: (Human) P78562

          View more View less

          Entrez Gene ID: (Human) 5251

          View more View less

          Function(s)
          metalloendopeptidase activity protein binding peptidase activity metallopeptidase activity zinc ion binding hydrolase activity metal ion binding
          Process(es)
          skeletal system development proteolysis cell-cell signaling protein processing organophosphate metabolic process bone mineralization lung development biomineral tissue development response to vitamin D protein modification process odontogenesis bone development response to growth hormone cellular response to vitamin D cellular response to parathyroid hormone stimulus response to sodium phosphate response to insulin-like growth factor stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline