Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MTRDEALPD SHSAQDFYEN YEPKEILGRG VSSVVRRCIH KPTSQEYAVK VIDVTGGGSF SPEEVRELRE ATLKEVDILR KVSGHPNIIQ LKDTYETNTF FFLVFDLMKR GELFDYLTEK VTLSEKETRK IMRALLEVIC TLHKLNIVHR DLKPENILLD DNMNIKLTDF GFSCQLEPGE RLREVCGTPS YLAPEIIECS MNEDHPGYGK EVDMWSTGVI MYTLLAGSPP FWHRKQMLML RMIMSGNYQF GSPEWDDYSD TVKDLVSRFL VVQPQNRYTA EEALAHPFFQ QYLVEEVRHF SPRGKFKVIA LTVLASVRIY YQYRRVKPVT REIVIRDPYA LRPLRRLIDA YAFRIYGHWV KKGQQQNRAA LFENTPKAVL LSLAEEDY (1-387 aa encoded by BC051327)
PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma phosphorylase kinase; PHK-gamma-M; PHK-gammaM; PHKgamma-M; PHKgammaM; phosphorylase kinase; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1; phosphorylase kinase, gamma 1 (muscle); serine/threonine-protein kinase PHKG1; unnamed protein product
Gene Aliases: PHKG; PHKIN01
UniProt ID: (Human) Q16816, (Mouse) P07934, (Rat) P13286
Entrez Gene ID: (Human) 5260, (Mouse) 18682, (Rat) 29353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support