Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTVEQKFGLF SAEIKEADPL AASEASQPKP CPPEVTPHYI WIDFLVQRFE IAKYCSSDQV EIFSSLLQRS MSLNIGRAKG SMNRHVAAIG PRFKLLTLGL SLLHADVVPN ATIRNVLREK IYSTAFDYFS CPPKFPTQGE KRLREDISIM IKFWTAMFSD KKYLTASQLV PPADIGDLLE QLVEENTGSL SGPAKDFYQR EFDFFNKITN VSAIIKPYPK GDERKKACLS ALSEVTVQPG CSLPSNPEAI VLDVDYKSGT PMQSAAKAPY LAKFKVKRCG VSELEKEGLR CRSDSEDECS TQEADGQKIS WQAAIFKLGD DCRQKSYWGA RTPTDRILRL PASQDMLALQ IIDLFKNIFQ LVGLDLFVFP YRVVATAPGC GVIECIPDCT SRDQLGRQTD FGMYDYFTRQ YGDESTLAFQ QACYNFIRSM AAYSLLLFLL QIKDRHNGNI MLDKKGHIIH IDFGFMFESS PGGNLGWEPD IKLTDEMVMI MGGKMEATPF KWFMEMCVQA TWLCGEPGPY MDVVVSLVTI MLDTGLPCFR GQTIKFLKHR FSPNMTEREA ANFIMKVIQS CFLSNRSRTY NMIQYYQNDI PY
Predicted to enable 1-phosphatidylinositol 4-kinase activity. Predicted to be involved in phosphatidylinositol phosphate biosynthetic process and phosphatidylinositol-mediated signaling. Predicted to be active in cytoplasm and plasma membrane. [provided by Alliance of Genome Resources, Apr 2022]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Em:AP000557.3; unnamed protein product
Entrez Gene ID: (Human) 375133
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support