Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PIAS1/PIAS2/PIAS3 Antibodies

          Abnova

          PIAS1/2/3 Recombinant Rabbit Monoclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PIAS1/2/3 Recombinant Rabbit Monoclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          RAB06469

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:100
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -

          Immunoprecipitation (IP)

          1:20
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          CHO cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          PIAS1, PIAS2, and PIAS3 are members of the Protein Inhibitor of Activated STAT (PIAS) family, which are integral to cellular processes such as SUMOylation, transcriptional regulation, and signal transduction. PIAS1 is noted for its role as a chromatin-bound androgen receptor (AR) coregulator, influencing AR-mediated target gene regulation in prostate cancer cells. It interacts with and modifies AR via SUMO3, which affects cell proliferation. PIAS2, another member, has been implicated in the inhibition of the JAK-STAT signaling pathway, notably suppressing STAT4-dependent gene induction linked to IL-12. Furthermore, PIAS2 has been shown to repress IFN-B production, highlighting its regulatory role in immune response mechanisms. PIAS3 is recognized as a SUMO E3 ligase that binds activated STAT3 and inhibits its DNA binding ability, subsequently hindering STAT3-mediated gene activation. Elevated expression of PIAS3 in prostate cancer samples has been associated with apoptosis induction. Together, the PIAS family genes are increasingly acknowledged in cancer biology due to their contributions to growth regulation and metastasis, while also representing potential therapeutic targets in cancer treatment strategies.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Androgen receptor-interacting protein 3; AR interacting protein; ARIP3; DAB2-interacting protein; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; DIP; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein ligase PIAS3; E3 SUMO-protein transferase PIAS1; E3 SUMO-protein transferase PIAS2; E3 SUMO-protein transferase PIAS3; GBP; Gu-binding protein; KChAP; Miz1; Msx-interacting zinc finger protein; Msx-interacting-zinc finger; Msx-interacting-zinc finger protein; nuclear protein that binds to nucleolar RNA helicase Gu protein and to p53; PIAS-NY protein; PIAS1/GBP; PIAS3; PIASxalpha6; PIG23; potassium channel regulatory protein KChAP; Potassium channel-associated protein; Protein inhibitor of activated STAT protein 1; Protein inhibitor of activated STAT protein 3; Protein inhibitor of activated STAT x; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS1; RING-type E3 ubiquitin transferase PIAS2; RNA helicase II-binding protein; unnamed protein product; zinc finger, MIZ-type containing 3; zinc finger, MIZ-type containing 4; zinc finger, MIZ-type containing 5

          View more View less

          Gene Aliases: 2900068C24Rik; 6330408K17Rik; AI462206; ARIP3; AU018068; DDXBP1; Dib; DIP; GBP; GU/RH-II; MIZ1; PIAS1; PIAS2; PIAS3; Pias3l; PIASX; PIASxalpha; PIASxb; PIASxbeta; SIZ2; ZMIZ3; ZMIZ4; ZMIZ5

          View more View less

          UniProt ID: (Human) O75925, (Human) O75928, (Human) Q9Y6X2, (Mouse) O88907, (Rat) Q6AZ28, (Mouse) Q8C5D8, (Mouse) O54714, (Rat) O70260

          View more View less

          Entrez Gene ID: (Human) 8554, (Human) 9063, (Human) 10401, (Mouse) 56469, (Rat) 300772, (Rat) 83422, (Mouse) 17344, (Mouse) 229615, (Rat) 83614

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding DNA binding transcription cofactor activity transcription corepressor activity protein binding zinc ion binding transferase activity SUMO transferase activity enzyme binding protein domain specific binding ubiquitin protein ligase binding metal ion binding SUMO ligase activity DNA-binding transcription factor binding RNA polymerase II sequence-specific DNA binding transcription factor binding protein C-terminus binding transcription factor binding UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity ligase activity ribosomal S6-glutamic acid ligase activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity estrogen receptor binding glucocorticoid receptor binding androgen receptor binding potassium channel regulator activity protein N-terminus binding
          Process(es)
          transcription, DNA-templated regulation of transcription, DNA-templated positive regulation of gene expression protein sumoylation positive regulation of protein sumoylation positive regulation of membrane potential regulation of transcription from RNA polymerase II promoter spermatogenesis metabolic process response to organic substance androgen receptor signaling pathway response to testosterone negative regulation of sequence-specific DNA binding transcription factor activity regulation of osteoblast differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of dendrite morphogenesis regulation of androgen receptor signaling pathway negative regulation of androgen receptor signaling pathway transforming growth factor beta receptor signaling pathway endoplasmic reticulum unfolded protein response negative regulation of transcription, DNA-templated regulation of macromolecule metabolic process SMAD protein signal transduction G1/S transition of mitotic cell cycle negative regulation of transcription from RNA polymerase II promoter JAK-STAT cascade positive regulation of proteasomal ubiquitin-dependent protein catabolic process regulation of cell proliferation negative regulation of apoptotic process fat cell differentiation positive regulation of smooth muscle cell differentiation protein-DNA complex assembly negative regulation of nucleic acid-templated transcription double-strand break repair via homologous recombination DNA double-strand break processing cellular response to DNA damage stimulus visual learning positive regulation of protein localization to cell periphery response to hormone negative regulation of gene expression regulation of tumor necrosis factor-mediated signaling pathway negative regulation of protein sumoylation negative regulation of osteoclast differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline