Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CALM Antibodies

          Abnova

          PICALM Polyclonal Antibody, MaxPab™

          View all (18) CALM antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PICALM Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PICALM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PICALM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PICALM Antibody (H00008301-B01P) in WB

          Western Blot analysis of PICALM expression in transfected 293T cell line by PICALM MaxPab polyclonal antibody. Lane 1: PICALM transfected lysate(67.1 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PICALM Antibody in Western Blot (WB)
          PICALM Polyclonal Antibody, MaxPab™

          Product Details

          H00008301-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PICALM (NP_001008660.1, 1 a.a. approximately 610 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.35 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSGQSLTDRI TAAQHSVTGS AVSKTVCKAT THEIMGPKKK HLDYLIQCTN EMNVNIPQLA DSLFERTTNS SWVVVFKSLI TTHHLMVYGN ERFIQYLASR NTLFNLSNFL DKSGLQGYDM STFIRRYSRY LNEKAVSYRQ VAFDFTKVKR GADGVMRTMN TEKLLKTVPI IQNQMDALLD FNVNSNELTN GVINAAFMLL FKDAIRLFAA YNEGIINLLE KYFDMKKNQC KEGLDIYKKF LTRMTRISEF LKVAEQVGID RGDIPDLSQA PSSLLDALEQ HLASLEGKKI KDSTAASRAT TLSNAVSSLA STGLSLTKVD EREKQAALEE EQARLKALKE QRLKELAKKP HTSLTTAASP VSTSAGGIMT APAIDIFSTP SSSNSTSKLP NDLLDLQQPT FHPSVHPMST ASQVASTWGG FTPSPVAQPH PSAGLNVDFE SVFGNKSTNV IVDSGGFDEL GGLLKPTVAS QNQNLPVAKL PPSKLVSDDL DSSLANLVGN LGIGNGTTKN DVNWSQPGEK KLTGGSNWQP KVAPTTAWNA ATMNGMHFPQ YAPPVMAYPA TTPTGMIGYG IPPQMGSVPV MTQPTLIYSQ PVMRPPNPFG PVSGAQIQFM

          Target Information

          PICALM, also known as the clathrin assembly lymphoid myeloid leukemia (CALM) protein, co-localizes with clathrin at clathrin-coated areas of the plasma membrane and in the Golgi area, and is thought to be an important component of the coated pit internalization machinery. Fusion proteins involving PICALM have been implicated in acute myeloid and T-cell acute lymphoblastic leukemia. PICALM is also expressed in neurons and is present at the synapse where it facilitates the endocytosis of the synaptic vesicle protein VAMP2. Recent studies have identified variants of PICALM associated with Alzheimer's disease and have suggested this protein may play a role in the endocytosis and recycling of the amyloid protein precursor (APP).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Clathrin assembly lymphoid myeloid leukemia protein; Phosphatidylinositol-binding clathrin assembly protein; unnamed protein product

          View more View less

          Gene Aliases: CALM; CLTH; LAP; PICALM

          View more View less

          UniProt ID: (Human) Q13492

          View more View less

          Entrez Gene ID: (Human) 8301

          View more View less

          Function(s)
          SNARE binding beta-amyloid binding protein binding phospholipid binding 1-phosphatidylinositol binding phosphatidylinositol-4,5-bisphosphate binding clathrin binding small GTPase binding clathrin heavy chain binding cadherin binding tau protein binding low-density lipoprotein particle receptor binding
          Process(es)
          cellular iron ion homeostasis endocytosis receptor-mediated endocytosis membrane budding axonogenesis learning or memory negative regulation of gene expression synaptic vesicle maturation vesicle-mediated transport endosomal transport hemopoiesis regulation of endocytosis receptor internalization regulation of protein localization cargo loading into vesicle positive regulation of transcription, DNA-templated positive regulation of Ras protein signal transduction regulation of receptor-mediated endocytosis negative regulation of receptor-mediated endocytosis clathrin coat assembly dendrite morphogenesis multicellular organismal iron ion homeostasis protein-containing complex assembly clathrin-mediated endocytosis regulation of vesicle size membrane bending amyloid-beta clearance by transcytosis regulation of beta-amyloid formation positive regulation of beta-amyloid formation regulation of amyloid precursor protein catabolic process positive regulation of amyloid precursor protein catabolic process negative regulation of protein localization to plasma membrane negative regulation of protein localization to cell surface
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline