Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PIGR Antibodies

          Proteintech

          PIGR Polyclonal Antibody, CoraLite® Plus 488

          View all (19) PIGR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PIGR Polyclonal Antibody, CoraLite® Plus 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          PIGR Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          PIGR Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PIGR Antibody (CL488-22024) in IHC (PFA)

          Immunofluorescent analysis of (4% PFA) fixed paraffin-embedded mouse small intestine tissue using CoraLite® Plus 488 PIGR antibody (CL488-22024) at dilution of 1:200. Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PIGR Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          PIGR Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          PIGR Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))

          Product Details

          CL488-22024

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PIGR fusion protein Ag16876

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: RAPAFEGRI LLNPQDKDGS FSVVITGLRK EDAGRYLCGA HSDGQLQEGS PIQAWQLFVN EESTIPRSPT VVKGVAGGSV AVLCPYNRKE SKSIKYWCLW EGAQNGRCPL LVDSEGWVKA QYEGRLSLLE EPGNGTFTVI LNQLTSRDAG FYWCLTNGDT LWRTTVEIKI IEGEPNLKVP GNVTAVLGET LKVPCHFPCK FSSYEKYWCK WNNTGCQALP SQDEGPSKAF VNCDENSRLV SLTLNLVTRA DEGWYWCGVK QGHFYGETAA VYVAVEERKA AGSRDVSLAK ADAAPDEKVL DSGFREIENK AIQDPRLFAE EKAVADTRDQ ADGSRASVDS GSSEEQGGSS R

          Target Information

          Polymeric IgA and IgM is produced and secreted by B cells in the lamina propria, which is beneath the mucosal lining of polarized epithelial cells. Polymeric immunoglobulin receptors, also designated pIgRs, are expressed on the basolateral surface of glandular epithelia and mediate transcellular transport of secretory immunoglobulin polymers across the epithelium. pIgR associates with secreted dimeric IgA and IgM molecules. During transcellular transport of these Ig polymers, pIgR undergoes proteolytic cleavage to generate a fragment called secretory component (SC), polymeric immunoglobulin receptor or poly-IG receptor. When immunoglobulin polymers associate with SC, they become resistant to enzymatic degradation during the transcytosis process. SC and the pIgR are crucial for proper mucosal immunity where they represent a molecular chaperone for polymeric Igs to remain intact and enter into body fluids. The human SC (pIgR) gene maps to chromosome 1q31-q41 and encodes a 764 amino acid protein. The receptor contains five units with homology to the variable (V) units of immunoglobulins and a transmembrane region that shares homology to certain immunoglobulin variable regions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hepatocellular carcinoma associated protein TB6; Hepatocellular carcinoma-associated protein TB6; phosphatidylinositol glycan, class R; PIgR; Poly Ig receptor; poly-Ig receptor; Polymeric immunoglobulin receptor; tb6

          View more View less

          Gene Aliases: PIGR

          View more View less

          UniProt ID: (Human) P01833, (Mouse) O70570

          View more View less

          Entrez Gene ID: (Human) 5284, (Mouse) 18703

          View more View less

          Function(s)
          polymeric immunoglobulin binding polymeric immunoglobulin receptor activity transmembrane signaling receptor activity epidermal growth factor receptor binding immunoglobulin receptor superfamily
          Process(es)
          detection of chemical stimulus involved in sensory perception of bitter taste immunoglobulin transcytosis in epithelial cells mediated by polymeric immunoglobulin receptor signal transduction epidermal growth factor receptor signaling pathway Fc receptor signaling pathway receptor clustering
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline