Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PIWIL4 Antibodies

          Abnova

          PIWIL4 Polyclonal Antibody, MaxPab™

          View all (12) PIWIL4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PIWIL4 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PIWIL4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PIWIL4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PIWIL4 Antibody (H00143689-B02P) in WB

          Western Blot analysis of PIWIL4 expression in transfected 293T cell line by PIWIL4 MaxPab polyclonal antibody. Lane 1: PIWIL4 transfected lysate(93.72 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PIWIL4 Antibody in Western Blot (WB)
          PIWIL4 Polyclonal Antibody, MaxPab™

          Product Details

          H00143689-B02P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PIWIL4 (ABM85351.1, 1 a.a. approximately 852 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSGRARVKAR GIARSPSATE VGRIQASPLP RSVDLSNNEA SSSNGFLGTS RISTNDKYGI SSGDAGSTFM ERGVKNKQDF MDLSICTREK LAHVRNCKTG SSGIPVKLVT NLFNLDFPQD WQLYQYHVTY IPDLASRRLR IALLYSHSEL SNKAKAFDGA ILFLSQKLEE KVTELSSETQ RGETIKMTIT LKRELPSSSP VCIQVFNIIF RKILKKLSMY QIGRNFYNPS EPMEIPQHKL SLWPGFAISV SYFERKLLFS ADVSYKVLRN ETVLEFMTAL CQRTGLSCFT QTCEKQLIGL IVLTRYNNRT YSIDDIDWSV KPTHTFQKRD GTEITYVDYY KQQYDITVSD LNQPMLVSLL KKKRNDNSEA QLAHLIPELC FLTGLTDQAT SDFQLMKAVA EKTRLSPSGR QQRLARLVDN IQRNTNARFE LETWGLHFGS QISLTGRIVP SEKILMQDHI CQPVSAADWS KDIRTCKILN AQSLNTWLIL CSDRTEYVAE SFLNCLRRVA GSMGFNVDYP KIIKVQENPA AFVRAIQQYV DPDVQLVMCI LPSNQKTYYD SIKKYLSSDC PVPSQCVLAR TLNKQGMMMS IATKIAMQMT CKLGGELWAV EIPLKSLMVV GIDVCKDALS KDVMVVGCVA SVNPRITRWF SRCILQRTMT DVADCLKVFM TGALNKWYKY NHDLPARIIV YRAGVGDGQL KTLIEYEVPQ LLSSVAESSS NTSSRLSVIV VRKKCMPRFF TEMNRTVQNP PLGTVVDSEA TRNEWYDFYL ISQVACRGTV SPTYYNVIYD DNGLKPDHMQ RLTFKLCHLY YNWPGIVSVP APCQYAHKLT FLVAQSIHKE PSLELANHLF YL

          Target Information

          PIWIL4 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: piwi-like 4; Piwi-like protein 4; PIWIL1; testis tissue sperm-binding protein Li 85P; unnamed protein product

          View more View less

          Gene Aliases: HIWI2; MIWI2; PIWI; PIWIL4

          View more View less

          UniProt ID: (Human) Q7Z3Z4

          View more View less

          Entrez Gene ID: (Human) 143689

          View more View less

          Function(s)
          nucleic acid binding RNA binding endoribonuclease activity piRNA binding
          Process(es)
          regulation of translation spermatogenesis epithelial structure maintenance cell differentiation gene silencing by RNA piRNA metabolic process meiotic cell cycle secondary piRNA processing transposable element silencing by heterochromatin formation transposable element silencing by piRNA-mediated DNA methylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline