Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PLSCR1 Antibodies

          Invitrogen

          PLSCR1 Polyclonal Antibody

          Advanced Verification
          View all (14) PLSCR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PLSCR1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Advanced Verification (2)
          PLSCR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PLSCR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PLSCR1 Antibody (PA5-110182) in WB

          Western blot was performed using Anti-PLSCR1 Polyclonal Antibody (Product # PA5-110182) and a 35 kDa band corresponding to Phospholipid scramblase 1 was observed across positive models (THP 1, HT-29, HaCaT, HCT 116 and Hep G2); and not across negative models (SW480 and SH-SY5Y). Membrane enriched extracts (30 µg lysate) of THP-1 (Lane 1), HT-29 (Lane 2), SW480 (Lane 3), HaCaT (Lane 4), HCT 116 (Lane 5), Hep G2 (Lane 6) and SH-SY5Y (Lane 7) were electrophoresed using NuPAGE™ 10% Bis-Tris Protein Gel (Product # NP0301BOX). Resolved proteins ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PLSCR1 Antibody in Western Blot (WB)
          PLSCR1 Antibody in Western Blot (WB)
          PLSCR1 Antibody in Western Blot (WB)
          PLSCR1 Antibody
          PLSCR1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PLSCR1 Polyclonal Antibody

          Product Details

          PA5-110182

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-318 of human Phospholipid Scramblase 1 (Phospholipid Scramblase 1 (PLSCR1)) (NP_0669281)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.59 mg/mL

          Amount

          59 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2855593

          Product Specific Information

          Immunogen sequence: MDKQNSQMNA SHPETNLPVG YPPQYPPTAF QGPPGYSGYP GPQVSYPPPP AGHSGPGPAG FPVPNQPVYN QPVYNQPVGA AGVPWMPAPQ PPLNCPPGLE YLSQIDQILI HQQIELLEVL TGFETNNKYE IKNSFGQRVY FAAEDTDCCT RNCCGPSRPF TLRIIDNMGQ EVITLERPLR CSSCCCPCCL QEIEIQAPPG VPIGYVIQTW HPCLPKFTIQ NEKREDVLKI SGPCVVCSCC GDVDFEIKSL DEQCVVGKIS KHWTGILREA FTDADNFGIQ FPLDLDVKMK AVMIGACFLI DFMFFESTGS QEQKSGVW

          Target Information

          The calcium-dependent plasma membrane protein phospholipid scramblase 1 (PLSCR1) contributes to the transbilayer movement of phosphatidylserine and other membrane phospholipids upon influx of calcium into the cytosol. This movement results in plasma membrane phospholipid remodelling and surface exposure of phosphatidylserine in injured or apoptotic cells, which leads to cell death. Interferons and other cytokines induce expression of PLSCR1, implying that PLSCR1 also functions in cytokine signaling pathways. EGF stilmulation results in tyrosine phosphorylation of PLSCR1 on tyrosines 69 and 74, which allows it to interact with Shc, and thereby connecting Src kinase activation to stimulation of the EGF receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ca(2+)-dependent phospholipid scramblase 1; erythrocyte phospholipid scramblase; hMmTRA1b; mg(2+)-dependent nuclease; PL scramblase 1; unnamed protein product

          View more View less

          Gene Aliases: MMTRA1B

          View more View less

          UniProt ID: (Human) O15162

          View more View less

          Entrez Gene ID: (Human) 5359

          View more View less

          Function(s)
          magnesium ion binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding virus receptor activity nuclease activity epidermal growth factor receptor binding calcium ion binding protein binding zinc ion binding SH3 domain binding phospholipid scramblase activity enzyme binding lead ion binding CD4 receptor binding mercury ion binding
          Process(es)
          phosphatidylserine biosynthetic process apoptotic process acute-phase response response to lead ion positive regulation of gene expression phospholipid scrambling platelet activation regulation of mast cell activation response to interferon-beta negative regulation of viral genome replication positive regulation of innate immune response positive regulation of transcription from RNA polymerase II promoter viral entry into host cell negative regulation of phagocytosis defense response to virus regulation of Fc receptor mediated stimulatory signaling pathway phosphatidylserine exposure on apoptotic cell surface positive regulation of chromosome separation positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0