Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PNLIPRP2 Antibodies

          Abnova

          PNLIPRP2 Polyclonal Antibody, MaxPab™

          View all (6) PNLIPRP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PNLIPRP2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          PNLIPRP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PNLIPRP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PNLIPRP2 Antibody (H00005408-D01P) in WB

          Western Blot analysis of PNLIPRP2 expression in transfected 293T cell line (H00005408-T01) by PNLIPRP2 MaxPab polyclonal antibody.

          Lane 1: PNLIPRP2 transfected lysate(51.90 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PNLIPRP2 Antibody in Western Blot (WB)
          PNLIPRP2 Antibody in Western Blot (WB)
          PNLIPRP2 Antibody in Western Blot (WB)
          PNLIPRP2 Polyclonal Antibody, MaxPab™

          Product Details

          H00005408-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PNLIPRP2 (AAH05989.1, 1 a.a. approximately 469 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MLPPWTLGLL LLATVRGKEV CYGQLGCFSD EKPWAGTLQR PVKLLPWSPE DIDTRFLLYT NENPNNFQLI TGTEPDTIEA SNFQLDRKTR FIIHGFLDKA EDSWPSDMCK KMFEVEKVNC ICVDWRHGSR AMYTQAVQNI RVVGAETAFL IQALSTQLGY SLEDVHVIGH SLGAHTAAEA GRRLGGRVGR ITGLDPAGPC FQDEPEEVRL DPSDAVFVDV IHTDSSPIVP SLGFGMSQKV GHLDFFPNGG KEMPGCKKNV LSTITDIDGI WEGIGGFVSC NHLRSFEYYS SSVLNPDGFL GYPCASYDEF QESKCFPCPA EGCPKMGHYA DQFKGKTSAV EQTFFLNTGE SGNFTSWRYK VSVTLSGKEK VNGYIRIALY GSNENSKQYE IFKGSLKPDA SHTCAIDVDF NVGKIQKVKF LWNKRGINLS EPKLGASQIT VQSGEDGTEY NFCSSDTVEE NVLQSLYPC

          Target Information

          PNLIPRP2 is a lipase with broad substrate specificity. It can hydrolyze both phospholipids and galactolipids. It acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cytotoxic T lymphocyte lipase; cytotoxic T-lymphocyte lipase; Galactolipase; Pancreatic lipase-related protein 2; PL-RP2; PNLIPRP2; Triacylglycerol lipase; unnamed protein product

          View more View less

          Gene Aliases: PLRP2; PNLIPRP2

          View more View less

          UniProt ID: (Human) P54317, (Mouse) P17892

          View more View less

          Entrez Gene ID: (Human) 5408, (Mouse) 18947

          View more View less

          Function(s)
          lipoprotein lipase activity phospholipase activity triglyceride lipase activity calcium ion binding protein binding phosphatidylcholine 1-acylhydrolase activity lipase activity hydrolase activity metal ion binding acylglycerol lipase activity galactolipase activity carboxylic ester hydrolase activity lipase
          Process(es)
          lipid metabolic process fatty acid biosynthetic process triglyceride metabolic process phospholipid catabolic process lipid catabolic process galactolipid catabolic process triglyceride catabolic process high-density lipoprotein particle remodeling phosphatidylcholine catabolic process cholesterol homeostasis lipid digestion cellular defense response digestion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline