Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • POLRMT Antibodies

          Abnova

          POLRMT Polyclonal Antibody, MaxPab™

          View all (9) POLRMT antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite POLRMT Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          POLRMT Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          POLRMT Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          POLRMT Antibody (H00005442-B01P) in WB

          Western Blot analysis of POLRMT expression in transfected 293T cell line by POLRMT MaxPab polyclonal antibody. Lane 1: POLRMT transfected lysate(135.3 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          POLRMT Antibody in Western Blot (WB)

          Product Details

          H00005442-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          POLRMT (AAH98387.1, 1 a.a. approximately 1230 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSALCWGRGA AGLKRALRPC GRPGLPGKEG TAGGVCGPRR SSSASPQEQD QDRRKDWGHV ELLEVLQARV RQLQAESVSE VVVNRVDVAR LPECGSGDGS LQPPRKVQMG AKDATPVPCG RWAKILEKDK RTQQMRMQRL KAKLQMPFQS GEFKALTRRL QVEPRLLSKQ MAGCLEDCTR QAPESPWEEQ LAQLLQEAPG KLSLDVEQAP SGQHSQAQLS GQQQRLLAFF KCCLLTDQLP LAHHLLVVHH GQRQKRKLLT LDMYNAVMLG WARQGAFKEL VYVLFMVKDA GLTPDLLSYA AALQCMGRQD QDAGTIERCL EQMSQEGLKL QALFTAVLLS EEDRATVLKA VHKVKPTFSL PPQLPPPVNT SKLLRDVYAK DGRVSYPKLH LPLKTLQCLF EKQLHMELAS RVCVVSVEKP TLPSKEVKHA RKTLKTLRDQ WEKALCRALR ETKNRLEREV YEGRFSLYPF LCLLDEREVV RMLLQVLQAL PAQGESFTTL ARELSARTFS RHVVQRQRVS GQVQALQNHY RKYLCLLASD AEVPEPCLPR QYWEALGAPE ALREQPWPLP VQMELGKLLA EMLVQATQMP CSLDKPHHSS RLVPVLYHVY SFRNVQQIGI LKPHPAYVQL LEKAAEPTLT FEAVDVPMLC PPLPWTSPHS GAFLLSPTKL MRTVEGATQH QELLETCPPT ALHGALDALT QLGNCAWRVN GRVLDLVLQL FQAKGCPQLG VPAPPSEAPQ PPEAHLPHSA APARKAELRR ELAHCQKVAR EMHSLRAEAL YRLSLAQHLR DRVFWLPHNM DFRGRTYPCP PHFNHLGSDV ARALLEFAQG RPLGPHGLDW LKIHLVNLTG LKKREPLRKR LAFAEEVMDD ILDSADQPLT GRKWWMGAEE PWQTLACCME VANAVRASDP AAYVSHLPVH QDGSCNGLQH YAALGRDSVG AASVNLEPSD VPQDVYSGVA AQVEVFRRQD AQRGMRVAQV LEGFITRKVV KQTVMTVVYG VTRYGGRLQI EKRLRELSDF PQEFVWEASH YLVRQVFKSL QEMFSGTRAI QHWLTESARL ISHMGSVVEW VTPLGVPVIQ PYRLDSKVKQ IGGGIQSITY THNGDISRKP NTRKQKNGFP PNFIHSLDSS HMMLTALHCY RKGLTFVSVH DCYWTHAADV SVMNQVCREQ FVRLHSEPIL QDLSRFLVKR FCSEPQKILE ASQLKETLQA VPKPGAFDLE QVKRSTYFFS

          Target Information

          This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DNA-directed RNA polymerase, mitochondrial; MtRPOL; polymerase (RNA) mitochondrial (DNA directed); unnamed protein product

          View more View less

          Gene Aliases: APOLMT; COXPD55; h-mtRPOL; MTRNAP; MTRPOL; POLRMT

          View more View less

          UniProt ID: (Human) O00411

          View more View less

          Entrez Gene ID: (Human) 5442

          View more View less

          Function(s)
          3'-5'-exoribonuclease activity mitochondrial RNA polymerase regulatory region DNA binding DNA binding RNA binding DNA-directed RNA polymerase activity protein binding transferase activity nucleotidyltransferase activity RNA polymerase activity sequence-specific DNA binding
          Process(es)
          mitochondrial DNA replication transcription, DNA-templated transcription from mitochondrial promoter transcription initiation from mitochondrial promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline