Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • POM121 Antibodies

          Proteintech

          POM121 Polyclonal Antibody

          8 References
          View all (6) POM121 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite POM121 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (13)
          POM121 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          POM121 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          POM121 Antibody (15645-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 15645-1-AP (POM121 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          POM121 Antibody in Immunocytochemistry (ICC/IF)
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          POM121 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          POM121 Antibody in Western Blot (WB)
          POM121 Antibody in Western Blot (WB)
          POM121 Antibody in Western Blot (WB)
          POM121 Antibody in Western Blot (WB)
          POM121 Polyclonal Antibody

          Product Details

          15645-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:10-1:100
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          POM121 Fusion Protein Ag8177 (635-984 aa encoded by BC008794)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.29 mg/mL

          Amount

          43.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          15645-1-AP do well in identifying the nuclear envelope in immunofluorescence experiments.

          Immunogen sequence: SQSLHTAVP TATSSSAADF SGFGSTLATS APATSSQPTL TFSNTSTPTF NIPFGSSAKS PLPSYPGANP QPAFGAAEGQ PPGAAKPALA PSFGSSFTFG NSAAPAAAPT PAPPSMIKVV PAYVPTPIHP IFGGATHSAF GLKATASAFG APASSQPAFG GSTAVFFGAA TSSGFGATTQ TASSGSSSSV FGSTTPSPFT FGGSAAPAGS GSFGINVATP GSSTTTGAFS FGAGQSGSTA TSTPFAGGLG QNALGTTGQS TPFAFNVSST TESKPVFGGT ATPTFGLNTP APGVGTSGSS LSFGASSAPA QGFVGVAPFG SAALSFSIGA GSKTPGARQR LQARRQHTRK K (635-984 aa encoded by BC008794)

          Target Information

          POM121 (POM121 membrane glycoprotein), also known as POM121A (nuclear envelope pore membrane protein POM 121A), NUP121 (nucleoporin Nup121) or pore membrane protein of 121 kDa, is a 1,249 amino acid single-pass membrane protein that exists as three alternatively spiced isoforms and belongs to the POM121 family. Encoded by a gene that maps to human chromosome 7q11.23, POM121 is highly conserved and localizes to nucleus membrane and endoplasmic reticulum membrane. An essential component of the nuclear pore complex (NPC), POM121 is mandatory for nuclear envelope formation and may play a role in biogenesis of the NPC. POM121 associates with the central spoke ring complex and is involved in anchoring components of the pore complex to the pore membrane by way of its F-X-F-G repeat-containing domain. POM121 overexpression induces the formation of cytoplasmic annulate lamellae (AL).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Nuclear envelope pore membrane protein POM 121; Nuclear envelope pore membrane protein POM 121A; nuclear pore membrane glycoprotein 121 kD; nuclear pore membrane protein 121; nuclear pore membrane protein 121 kDa; nuclear pore membrane protein 121-2; Nucleoporin Nup121; P145; POM121 membrane glycoprotein; POM121 membrane glycoprotein C; POM121B; Pore membrane protein of 121 kDa; pore membrane protein of 121 kDa C; unnamed protein product

          View more View less

          Gene Aliases: 2610027A18Rik; C80273; KIAA0618; mKIAA0618; NUP121; P145; POM121; POM121-2; POM121A

          View more View less

          UniProt ID: (Human) Q96HA1, (Rat) P52591, (Mouse) Q8K3Z9

          View more View less

          Entrez Gene ID: (Human) 9883, (Human) 100101267, (Rat) 113975, (Mouse) 107939

          View more View less

          Function(s)
          protein binding nuclear localization sequence binding structural constituent of nuclear pore transporter
          Process(es)
          RNA export from nucleus protein import into nucleus nucleocytoplasmic transport protein transport mRNA transport nuclear pore organization transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0