Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PP2A alpha Antibodies

          Invitrogen

          PP2A alpha Polyclonal Antibody

          Advanced Verification
          View all (27) PP2A alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PP2A alpha Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          • Advanced Verification (1)
          PP2A alpha Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PP2A alpha Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PP2A alpha Antibody (PA5-29754) in ICC/IF

          Immunofluorescence analysis of PP2A alpha was performed using 70% confluent log phase PC-3 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with PP2A alpha Polyclonal Antibody (Product # PA5-29754) at 1:500 dilution in 0.1% BSA, incubated at 4 degree celsiu... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PP2A alpha Antibody in Immunocytochemistry (ICC/IF)
          PP2A alpha Antibody in Immunocytochemistry (ICC/IF)
          PP2A alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PP2A alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PP2A alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PP2A alpha Antibody in Western Blot (WB)
          PP2A alpha Antibody in Western Blot (WB)
          PP2A alpha Antibody in Western Blot (WB)
          PP2A alpha Antibody in Western Blot (WB)
          PP2A alpha Antibody in Western Blot (WB)
          PP2A alpha Antibody

          Product Details

          PA5-29754

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human PP2A alpha. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.45 mg/mL

          Amount

          45 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 1% BSA, 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2547228

          Product Specific Information

          Recommended positive controls: HCT116, HeLa, HepG2, Molt-4, mouse brain.

          Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Xenopus laevis (100%), Dog (99%), Pig (100%), Rabbit (100%), Chicken (100%), Bovine (100%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. An additional protein phosphatase catalytic subunit, PPX (also known as PP4) is a putative member of a novel PP family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: B55-GAMMA; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; phosphatase 2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; PP2A-alpha; PPP2CA; PR52; PR55G; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase-2A-alpha; Replication protein C; RP-C; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product

          View more View less

          Gene Aliases: HJS3; NEDLBA; PP2A; Pp2a1; PP2Ac; PP2CA; PP2Calpha; PPP2CA; R75353; RP-C

          View more View less

          UniProt ID: (Human) P67775, (Mouse) P63330, (Rat) P63331

          View more View less

          Entrez Gene ID: (Human) 5515, (Mouse) 19052, (Rat) 24672

          View more View less

          Function(s)
          phosphoprotein phosphatase activity protein serine/threonine phosphatase activity protein tyrosine phosphatase activity protein binding hydrolase activity metal ion binding protein heterodimerization activity tau protein binding GABA receptor binding RNA polymerase II CTD heptapeptide repeat S2 phosphatase activity RNA polymerase II CTD heptapeptide repeat S5 phosphatase activity RNA polymerase II CTD heptapeptide repeat S7 phosphatase activity protein C-terminus binding enzyme binding protein kinase binding protein phosphatase binding protein domain specific binding protein complex binding protein kinase B binding ion channel binding protein dimerization activity protein phosphatase 2A binding protein antigen binding protein phosphatase
          Process(es)
          mitotic cell cycle regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter transcription elongation from RNA polymerase II promoter protein dephosphorylation mesoderm development response to lead ion negative regulation of epithelial to mesenchymal transition regulation of microtubule polymerization negative regulation of hippo signaling intracellular signal transduction peptidyl-threonine dephosphorylation regulation of growth T cell homeostasis regulation of cell differentiation meiotic cell cycle negative regulation of protein kinase B signaling negative regulation of canonical Wnt signaling pathway vascular endothelial cell response to oscillatory fluid shear stress RNA polymerase II transcription initiation surveillance positive regulation of NLRP3 inflammasome complex assembly negative regulation of glycolytic process through fructose-6-phosphate regulation of G1/S transition of mitotic cell cycle negative regulation of transcription from RNA polymerase II promoter regulation of protein phosphorylation negative regulation of protein phosphorylation regulation of receptor activity regulation of protein autophosphorylation positive regulation of protein phosphatase type 2A activity positive regulation of protein dephosphorylation regulation of protein catabolic process negative regulation of transcription factor import into nucleus positive regulation of apoptotic process positive regulation of cysteine-type endopeptidase activity involved in apoptotic process regulation of cell cycle protein heterotrimerization cellular response to glucose stimulus positive regulation of protein serine/threonine kinase activity negative regulation of calcium ion transmembrane transporter activity cardiac ventricle development heart development cellular response to insulin stimulus cellular response to calcium ion cellular response to cytokine stimulus cellular response to ethanol cellular response to follicle-stimulating hormone stimulus cellular response to steroid hormone stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline