Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PPAP2B Antibodies

          Proteintech

          PPAP2B Polyclonal Antibody

          View all (9) PPAP2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PPAP2B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PPAP2B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PPAP2B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPAP2B Antibody (27391-1-AP) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 27391-1-AP (PPAP2B antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPAP2B Antibody in Western Blot (WB)
          PPAP2B Polyclonal Antibody

          Product Details

          27391-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PPAP2B fusion protein Ag26422
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          60 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MQNYKYDKAI VPESKNGGSP ALNNNPRRSG SKRVLLICLD LFCLFMAGLP FLIIETSTIK PYHRGFYCND ESIKYPLKTG ETINDAVLCA VGIVIAILAI ITGEFYRIYY LKKSRSTIQN

          Target Information

          Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EC 3.1.3.4; Lipid phosphate phosphohydrolase 3; MGC15306; PAP-2b; PAP2 beta; PAP2-b; PAP2-beta; PAP2b; Phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; Phospholipid phosphatase 3; type-2 phosphatidic acid phosphatase-beta; unnamed protein product; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP

          View more View less

          Gene Aliases: 1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; Dri42; LPP3; PAP2B; PLPP3; Ppab2b; PPAP2B; VCIP

          View more View less

          UniProt ID: (Human) O14495, (Mouse) Q99JY8

          View more View less

          Entrez Gene ID: (Human) 8613, (Mouse) 67916

          View more View less

          Function(s)
          integrin binding protein binding phosphatidate phosphatase activity hydrolase activity sphingosine-1-phosphate phosphatase activity lipid phosphatase activity delta-catenin binding ceramide-1-phosphate phosphatase activity catalytic activity phosphatase
          Process(es)
          blood vessel development gastrulation with mouth forming second lipid metabolic process phospholipid metabolic process sphingosine metabolic process ceramide metabolic process retrograde vesicle-mediated transport, Golgi to ER cell adhesion signal transduction integrin-mediated signaling pathway positive regulation of endothelial cell migration regulation of Wnt signaling pathway sphingolipid catabolic process cell-cell adhesion mediated by integrin positive regulation of homotypic cell-cell adhesion positive regulation of transcription from RNA polymerase II promoter phospholipid dephosphorylation protein stabilization Bergmann glial cell differentiation cell-cell adhesion regulation of sphingolipid mediated signaling pathway positive regulation of intracellular signal transduction positive regulation of endothelial cell-matrix adhesion via fibronectin negative regulation of protein phosphorylation multicellular organism development dephosphorylation single organismal cell-cell adhesion homotypic cell-cell adhesion canonical Wnt signaling pathway involved in positive regulation of endothelial cell migration canonical Wnt signaling pathway involved in positive regulation of cell-cell adhesion canonical Wnt signaling pathway involved in positive regulation of wound healing positive regulation of peptidyl-tyrosine phosphorylation positive regulation of sequence-specific DNA binding transcription factor activity canonical Wnt signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0