Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAGNDCGAL LDEELSSFFL NYLADTQGGG SGEEQLYADF PELDLSQLDA SDFDSATCFG ELQWCPENSE TEPNQYSPDD SELFQIDSEN EALLAELTKT LDDIPEDDVG LAAFPALDGG DALSCTSASP APSSAPPSPA PEKPSAPAPE VDELSLLQKL LLATSYPTSS SDTQKEGTAW RQAGLRSKSQ RPCVKADSTQ DKKAPMMQSQ SRSCTELHKH LTSAQCCLQD RGLQPPCLQS PRLPAKEDKE PGEDCPSPQP APASPRDSLA LGRADPGAPV SQEDMQAMVQ LIRYMHTYCL PQRKLPPQTP EPLPKACSNP SQQVRSRPWS RHHSKASWAE FSILRELLAQ D (1-350 aa encoded by BC132971)
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERR ligand 1; PERC; PERC-s; lacks one estrogen receptor interaction domain; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; pgc-1 beta; PGC-1-related estrogen receptor alpha coactivator; PGC1beta-1a; PGC1beta-1b; PGC1beta-2a; PGC1beta-2b; PPAR gamma coactivator-1beta protein; PPAR-gamma coactivator 1-beta; PPARAGCIbeta; PPARgamma coactivator 1 beta; PPARGC-1-beta; unnamed protein product
Gene Aliases: 4631412G21Rik; ERRL1; PERC; PGC-1(beta); PGC-1beta; PGC-1beta/ERRL1; PGC1B; PPARGC-1-beta
UniProt ID: (Human) Q86YN6, (Mouse) Q8VHJ7
Entrez Gene ID: (Human) 133522, (Mouse) 170826
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support