Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TTEVPLALE EESPSEGCPS SEIPMEKEPG EGRISVVDYS YLEGDLPISA RPACSNKLID YILGGASSDL ETSSDPEGED WDEEAEDDGF DSDSSLSDSD LEQDPEGLHL WNSFCSVDPY NPQNFTATIQ TAARIVPEEP SDSEKDLSGK SDLENSSQSG SLPETPEHSS GEEDDWESSA DEAESLKLWN SFCNSDDPYN PLNFKAPFQT SGENEKGCRD SETPSESIVA ISECHTLLSC KVQLLGSQES ECPDSVQRDV LSGGRHTHVK RKKVTFLEEV TEYYISGDED RKGPWEEFAR DGCRFQKRIQ ETEDAIGYCL TFEHRERMFN RLQGTCFKGL NVLKQC (369-713 aa encoded by BC065280)
Maintains low levels of EIF2S1 phosphorylation in unstressed cells by promoting its dephosphorylation by PP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: constitutive repressor of eIF2alpha phosphorylation; protein phosphatase 1, regulatory (inhibitor) subunit 15B; unnamed protein product
Gene Aliases: 1810033K10Rik; C530022L24Rik; CREP; MSSGM2
UniProt ID: (Human) Q5SWA1, (Mouse) Q8BFW3
Entrez Gene ID: (Human) 84919, (Mouse) 108954, (Rat) 304799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support