Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • PPP1R9B Antibodies

          Invitrogen

          PPP1R9B Polyclonal Antibody

          Advanced Verification
          View all (7) PPP1R9B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PPP1R9B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Advanced Verification (1)
          PPP1R9B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PPP1R9B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPP1R9B Antibody (PA5-48102) in ICC/IF

          Immunofluorescence analysis of PPP1R9B was performed using MCF7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with PPP1R9B Sheep Polyclonal Antibody (Product # PA5-48102) at 5 µg/mL in 0.1% BSA and incubated overnight at 4 degree and then labeled with Donkey anti-Sheep IgG (H+L) Cross-Adsorbed, Alexa Fluor® 488 conjugate secondary antibody (Product # A-11015) at a dilution of 1:2000 for... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPP1R9B Antibody in Immunocytochemistry (ICC/IF)
          PPP1R9B Antibody in Immunocytochemistry (ICC/IF)
          PPP1R9B Antibody in Western Blot (WB)
          PPP1R9B Antibody in Western Blot (WB)
          PPP1R9B Antibody in Western Blot (WB)
          PPP1R9B Antibody
          PPP1R9B Polyclonal Antibody

          Product Details

          PA5-48102

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          5-15 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Sheep / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E. coli-derived recombinant human PPP1R9B Lys289-Asp412
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2605900

          Product Specific Information

          Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

          Target Information

          Protein phosphatase 1 regulatory subunit 9B (PPP1R9B), also known as Neurabin II or spinophilin, is a gene located on chromosome 17q21.32 that encodes a regulatory protein predominantly expressed in the brain. PPP1R9B is involved in modulating the activity of protein phosphatase 1 (PP1), a crucial enzyme in cellular signaling pathways responsible for dephosphorylation of target proteins. Spinophilin functions by anchoring PP1 to specific subcellular locations, such as synaptic sites, thereby influencing synaptic plasticity, neurotransmission, and signal transduction. This protein is integral to the maintenance and modulation of excitatory synapses, playing a role in cognitive processes including learning and memory. Alterations in PPP1R9B expression or function have been associated with neuropsychiatric disorders and cancer, reflecting its importance in neuronal integrity and its potential role in tumorigenesis. Research into PPP1R9B continues to explore its therapeutic potential in neurological disorders and its utility in understanding phosphatase-related cellular processes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: neurabin 2; Neurabin-2; Neurabin-II; Neural tissue-specific F-actin-binding protein II; nuerabin 2; p130; PP1bp134; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; Spinophilin; unnamed protein product

          View more View less

          Gene Aliases: Neb2; PPP1R6; PPP1R9; PPP1R9B; SPINO; Spn

          View more View less

          UniProt ID: (Human) Q96SB3, (Rat) O35274

          View more View less

          Entrez Gene ID: (Human) 84687, (Rat) 84686

          View more View less

          Function(s)
          actin binding protein kinase activity protein phosphatase inhibitor activity protein binding protein phosphatase 1 binding kinase binding D2 dopamine receptor binding ion channel binding macromolecular complex binding actin filament binding
          Process(es)
          regulation of protein phosphorylation actin filament organization dendrite development cell migration calcium-mediated signaling negative regulation of cell growth regulation of cell proliferation filopodium assembly modulation of synaptic transmission cellular response to morphine regulation of opioid receptor signaling pathway regulation of cell growth by extracellular stimulus response to amphetamine developmental process involved in reproduction regulation of exit from mitosis nervous system development learning RNA splicing response to xenobiotic stimulus hippocampus development cerebral cortex development actin filament depolymerization cell differentiation neuron projection development response to estradiol response to prostaglandin E response to nicotine response to immobilization stress response to amino acid response to steroid hormone regulation of cell cycle male mating behavior reproductive system development cellular response to epidermal growth factor stimulus cellular response to estradiol stimulus cellular response to xenobiotic stimulus regulation of postsynapse assembly cellular response to peptide positive regulation of protein localization to plasma membrane protein localization to actin cytoskeleton positive regulation of cellular protein localization positive regulation of protein localization to actin cortical patch response to kainic acid response to L-phenylalanine derivative protein localization to cell periphery
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline