Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PPP2R2B/A/C/D Antibodies

          Proteintech

          PPP2R2B/A/C/D Polyclonal Antibody

          2 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PPP2R2B/A/C/D Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          PPP2R2B/A/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PPP2R2B/A/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPP2R2B/A/C/D Antibody (29237-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using 29237-1-AP (PPP2R2B/A/C/D antibody) at dilution of 1:400 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPP2R2B/A/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPP2R2B/A/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PPP2R2B/A/C/D Antibody in Western Blot (WB)
          PPP2R2B/A/C/D Polyclonal Antibody

          Product Details

          29237-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:400-1:800
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PPP2R2B/A/C/D fusion protein Ag30757
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.6 mg/mL

          Amount

          90 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: SHSGRYIMTR DYLTVKVWDL NMENRPIETY QVHDYLRSKL CSLYENDCIF DKFECVWNGS DSVIMTGSYN NFFRMFDRNT KRDVTLEASR ENSKPRAILK PRKVCVGGKR RKDEISVDSL DFSKKILHTA WHPSENIIAV AATNNLYIFQ DKVN

          Target Information

          The genes PPP2R2B, PPP2R2A, PPP2R2C, and PPP2R2D encode different regulatory subunits of protein phosphatase 2 (PP2A), a major serine/threonine phosphatase implicated in various cellular processes. PPP2R2B is located on chromosome 5q32 and has multiple alternatively spliced transcripts, playing a significant role in cancer prognosis and immune evasion (ScienceDirect). PPP2R2A, encoding the B55 alpha subunit, is frequently deleted in prostate adenocarcinomas, correlating with poor prognosis (PubMed). PPP2R2C, encoding the B55 gamma isoform, is shown to suppress glioma cell proliferation, acting as a tumor suppressor (ScienceDirect). PPP2R2D is involved in T-cell regulation and cancer cell proliferation, with overexpression linked to increased mTOR activity and impact on IL-2 production (PubMed). Collectively, these genes highlight the complexity and critical regulatory roles of PP2A subunits in cell signaling and disease pathology.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B55; BRbeta B-regulatory subunit of protein phosphatase 2A; IMYPNO1; phosphoprotein phosphatase 2A BR gamma regulatory chain; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform B55-beta; PP2A subunit B isoform B55-delta; PP2A subunit B isoform B55-gamma; PP2A subunit B isoform beta; PP2A subunit B isoform BRA; PP2A subunit B isoform BRB; PP2A subunit B isoform BRgamma; PP2A subunit B isoform delta; PP2A subunit B isoform gamma; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform PR55-beta; PP2A subunit B isoform PR55-delta; PP2A subunit B isoform PR55-gamma; PP2A subunit B isoform R2-alpha; PP2A subunit B isoform R2-beta; PP2A subunit B isoform R2-delta; PP2A subunit B isoform R2-gamma; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, B-beta isoform; PP2A, subunit B, B-delta isoform; PP2A, subunit B, B-gamma isoform; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B55-beta isoform; PP2A, subunit B, B55-delta isoform; PP2A, subunit B, B55-gamma isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, BRB isoform; PP2A, subunit B, BRgamma isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, PR55-beta isoform; PP2A, subunit B, PR55-delta isoform; PP2A, subunit B, PR55-gamma isoform; PP2A, subunit B, R2-alpha isoform; PP2A, subunit B, R2-beta isoform; PP2A, subunit B, R2-delta isoform; PP2A, subunit B, R2-gamma isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gamma isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform; protein phosphatase 2 regulatory subunit B, delta; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2, regulatory subunit B, beta; protein phosphatase 2, regulatory subunit B, delta isoform; protein phosphatase 2A B regulatory subunit delta isoform; protein phosphatase 2A, regulatory subunit B, delta isoform; protein phosphatase 2A1 B gamma subunit; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gamma isoform; unnamed protein product; upstream ORF 1; upstream ORF 2

          View more View less

          Gene Aliases: 1300017E19Rik; 2410004D02Rik; 2900026H06Rik; 6330404L05Rik; 6330548O06Rik; B55; B55-GAMMA; B55A; B55ALPHA; B55BETA; B55D; B55delta; B55gamma; D7Ertd753e; E130009M08Rik; IMYPNO; IMYPNO1; KIAA1541; MDS026; mKIAA1541; PP2A-PR55B; PP2AB55BETA; PP2ABBETA; PP2APR55B; PP2APR55BETA; PPP2R2A; PPP2R2B; PPP2R2C; PPP2R2D; Pppr2b2; PR2AB55BETA; PR2ABBETA; PR2APR55BETA; PR52; PR52A; PR52B; PR55-BETA; PR55A; PR55alpha; PR55BETA; PR55G; SCA12

          View more View less

          UniProt ID: (Human) P63151, (Human) Q00005, (Human) Q9Y2T4, (Human) Q66LE6, (Mouse) Q6P1F6, (Rat) P36876, (Rat) P36877, (Mouse) Q6ZWR4, (Mouse) Q8BG02, (Rat) P97888, (Rat) P56932, (Mouse) Q925E7

          View more View less

          Entrez Gene ID: (Human) 5520, (Human) 5521, (Human) 5522, (Human) 55844, (Mouse) 71978, (Rat) 117104, (Rat) 60660, (Mouse) 72930, (Mouse) 269643, (Rat) 117256, (Rat) 246255, (Mouse) 52432

          View more View less

          Function(s)
          protein binding protein phosphatase regulator activity macromolecular complex binding tau protein binding protein phosphatase 2A binding enzyme-substrate adaptor activity protein serine/threonine phosphatase activity protein phosphatase type 2A regulator activity protein complex binding phosphatidate phosphatase activity protein phosphatase
          Process(es)
          protein dephosphorylation response to morphine regulation of chromosome segregation apoptotic process mitotic cell cycle exit from mitosis cell division signal transduction regulation of catalytic activity mitochondrial fission protein targeting to mitochondrion spermatid development brain development neuron differentiation regulation of protein phosphatase type 2A activity positive regulation of neuron apoptotic process mitochondrial fragmentation involved in apoptotic process mitotic nuclear division dephosphorylation cell cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0