Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LGIFSKTWIK AGTEMGPFTG RVIAPEHVDI CKNNNLMWEV FNEDGTVRYF IDASQEDHRS WMTYIKCARN EQEQNLEVVQ IGTSIFYKAI EMIPPDQELL VWYGNSHNTF LGIPGVPGLE EDQKKNKHED FHPADSAAGP AGRMRCVICH RGFNSRSNLR SHMRIHTLDK PFVCRFCNRR FSQSSTLRNH VRLHTGERPY KCQVCQSAYS QLAGLRAHQK SARHRPPSTA L
This gene encodes a transcriptional regulator of sensory neuronal specification that plays a critical role in pain perception. The encoded protein contains an N-terminal PRDI-BF1 and RIZ homology (PR) domain, a SET domain, and three C-terminal C2H2 zinc finger DNA-binding domains. Naturally occurring mutations in this gene are associated with congenital insensitivity to pain (CIP), and hereditary sensory and autonomic neuropathies (HSAN's) affecting peripheral sensory and autonomic neurons. Deregulation of this gene is associated with solid cancers and hematological malignancies including chronic myeloid leukemia.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PR domain containing 12; PR domain-containing protein 12; RP11-57C19.7
Gene Aliases: Gm998; HSAN8; PFM9
UniProt ID: (Human) Q9H4Q4, (Mouse) A2AJ77
Entrez Gene ID: (Human) 59335, (Mouse) 381359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support