Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PRDM16 Antibodies

          Invitrogen

          PRDM16 Polyclonal Antibody

          35 Published Figures
          26 References
          View all (10) PRDM16 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PRDM16 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (35)
          PRDM16 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          PRDM16 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 39

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PRDM16 Antibody (PA5-20872) in IHC

          Immunofluorescence of PRDM16 in mouse brain tissue with PRDM16 Polyclonal Antibody (Product # PA5-20872) at 20 µg/mL. Green: PRDM16 Blue: DAPI staining {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PRDM16 Antibody in Immunohistochemistry (IHC)
          PRDM16 Antibody in Immunohistochemistry (IHC)
          PRDM16 Antibody in Immunohistochemistry (IHC)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PRDM16 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PRDM16 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PRDM16 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot, ChIP Assay (WB, ChIP)
          PRDM16 Antibody in Western Blot, ChIP Assay (WB, ChIP)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)
          PRDM16 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PRDM16 Polyclonal Antibody

          Product Details

          PA5-20872

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          View 22 publications 22 publications

          Immunohistochemistry (IHC)

          2 µg/mL
          View 3 publications 3 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Rabbit, Sheep

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A 17 amino acid peptide from near the carboxy terminus of human PRDM16.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11154178

          Product Specific Information

          PA5-20872 can be used with blocking peptide PEP-0986.

          Target Information

          PRDM16 is a zinc finger transcription factor and contains an N-terminal PR domain. The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36, q21)-positive MDS/AML. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Recent studies have shown that PRDM16 normally acts as a Smad3 binding protein that may be important for the development of orofacial structures through modulation of the TGF-beta signaling pathway. Other experiments have indicated that PRDM16 controls a bidirectional cell fate switch between skeletal myoblasts and brown fat cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: line 27; MDS1/EVI1-like gene 1; PR domain 16; PR domain containing 16; PR domain zinc finger protein 16; PR domain-containing protein 16; PRDM16; PFM13; transcription factor MEL1; unnamed protein product

          View more View less

          Gene Aliases: 5730557K01Rik; CMD1LL; csp1; KMT8F; LVNC8; MEL1; PFM13

          View more View less

          UniProt ID: (Human) Q9HAZ2, (Mouse) A2A935

          View more View less

          Entrez Gene ID: (Human) 63976, (Mouse) 70673

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding transcription factor activity, sequence-specific DNA binding transcription cofactor activity transcription coactivator activity transcription corepressor activity protein binding sequence-specific DNA binding histone methyltransferase activity (H3-K9 specific) DNA-binding transcription factor binding chromatin-protein adaptor activity histone H3 methyltransferase activity histone H3K9 monomethyltransferase activity SMAD binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter tolerance induction in gut-associated lymphoid tissue chromatin remodeling regulation of transcription from RNA polymerase II promoter negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of granulocyte differentiation regulation of cellular respiration regulatory T cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter brown fat cell differentiation negative regulation of muscle cell differentiation protein maturation heterochromatin organization protein localization to chromatin positive regulation of cold-induced thermogenesis beige fat cell differentiation negative regulation of white fat cell differentiation immune system process stem cell population maintenance neurogenesis hemopoiesis somatic stem cell population maintenance tongue development white fat cell differentiation palate development positive regulation of brown fat cell differentiation maintenance of cell number
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-dg9d8:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0