Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MMLRGNLKQ VRIEKNPARL RALESAVGES EPAAAAAMAL ALAGEPAPPA PAPPEDHPDE EMGFTIDIKS FLKPGEKTYT QRCRLFVGNL PTDITEEDFK RLFERYGEPS EVFINRDRGF GFIRLESRTL AEIAKAELDG TILKSRPLRI RFATHGAALT VKNLSPVVSN ELLEQAFSQF GPVEKAVVVV DDRGRATGKG FVEFAAKPPA RKALERCGDG AFLLTTTPRP VIVEPMEQFD DEDGLPEKLM QKTQQYHKER EQPPRFAQPG TFEFEYASRW KALDEMEKQQ REQVDRNIRE AKEKLEAEME AARHEHQLML MRQDLMRRQE ELRRLEELRN QELQKRKQIQ LR (1-351 aa encoded by BC014184)
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: paraspeckle localization; paraspeckle protein 1; PSP1; PSPC1; unnamed protein product
Gene Aliases: 5730470C09Rik; PSP1
UniProt ID: (Human) Q8WXF1, (Mouse) Q8R326, (Rat) Q4KLH4
Entrez Gene ID: (Human) 55269, (Mouse) 66645, (Rat) 305910
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support