Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
PA1-4135 detects Patched 2 (PTCH2) in human and mouse samples.
PA1-4135 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing PTCH2 in fetal brain lysate.
The PA1-4135 immunogen is a synthetic peptide corresponding to residues G(226) P F A S L E G F R(235) of human PTCH2.
Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12 TM receptor protein; Hedgehog receptor; patched homolog 2; ptch2; RP11-269F19.8
Gene Aliases: PTC2; SLC65B2
UniProt ID: (Human) Q9Y6C5, (Mouse) O35595
Entrez Gene ID: (Human) 8643, (Mouse) 19207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support