Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PTPN22 Antibodies

          Abnova

          PTPN22 Polyclonal Antibody, MaxPab™

          View all (16) PTPN22 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PTPN22 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PTPN22 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PTPN22 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTPN22 Antibody (H00026191-B01P) in WB

          Western Blot analysis of PTPN22 expression in transfected 293T cell line by PTPN22 MaxPab polyclonal antibody. Lane1:PTPN22 transfected lysate(82.72 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTPN22 Antibody in Western Blot (WB)
          PTPN22 Polyclonal Antibody, MaxPab™

          Product Details

          H00026191-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PTPN22 (AAH71670.1, 1 a.a. approximately 752 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDQREILQKF LDEAQSKKIT KEEFANEFLK LKRQSTKYKA DKTYPTTVAE KPKNIKKNRY KDILPYDYSR VELSLITSDE DSSYINANFI KGVYGPKAYI ATQGPLSTTL LDFWRMIWEY SVLIIVMACM EYEMGKKKCE RYWAEPGEMQ LEFGPFSVSC EAEKRKSDYI IRTLKVKFNS ETRTIYQFHY KNWPDHDVPS SIDPILELIW DVRCYQEDDS VPICIHCSAG CGRTGVICAI DYTWMLLKDG SQAKHCIPEK NHTLQADSYS PNLPKSTTKA AKMMNQQRTK MEIKESSSFD FRTSEISAKE ELVLHPAKSS TSFDFLELNY SFDKNADTTM KWQTKAFPIV GEPLQKHQSL DLGSLLFEGC SNSKPVNAAG RYFNSKVPIT RTKSTPFELI QQRETKEVDS KENFSYLESQ PHDSCFVEMQ AQKVMHVSSA ELNYSLPYDS KHQIRNASNV KHHDSSALGV YSYIPLVENP YFSSWPPSGT SSKMSLDLPE KQDGTVFPSS LLPTSSTSLF SYYNSHDSLS LNSPTNISSL LNQESAVLAT APRIDDEIPP PLPVRTPESF IVVEEAGEFS PNVPKSLSSA VKVKIGTSLE WGGTSEPKKF DDSVILRPSK SVKLRSPKSE LHQDRSSPPP PLPERTLESF FLADEDCMQA QSIETYSTSY PDTMENSTSS KQTLKTPGKS FTRSKSLKIL RNMKKSICNS CPPNKPAESV QSNNSSSFLN FGFANRFSKP KGPRNPPPTW NI

          Target Information

          The protein tyrosine phosphatase PTPN22 (PTP22, LYP, PEP, formerly PTPN8) is a genetic variant that confers risk of developing diverse human autoimmune diseases such as type 1 diabetes and rheumatoid arthritis. The minor allele of a missense SNP in PTPN22 encodes a hematopoietic-specific protein tyrosine phosphatase also known as "Lyp." The risk allele is present in about 17% of Caucasian individuals from the general population and in approximately 28% of Caucasian individuals with rheumatoid arthritis; it is thought to disrupt the P1 proline-rich motif that is important for interaction with the Src homology-3 (SH3) domain of CSK (cytoplasmic tyrosine kinase), potentially altering the normal functions of these proteins as negative regulators of T cell activation. The interaction between CSK and PTPN22 is highly specific and it is speculated that PTPN22 may be an effector and/or regulator of CSK in T cells and other hematopoietic cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP; Lymphoid phosphatase; lymphoid-specific protein tyrosine phosphatase; LyP; LYP3; PEP; PEST-domain phosphatase; protein tyrosine phosphatase, non-receptor type 22 (lymphoid); protein tyrosine phosphatase, non-receptor type 8; PTPN22.8; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 22; unnamed protein product

          View more View less

          Gene Aliases: LYP; LYP1; LYP2; PEP; PTPN22; PTPN22.5; PTPN22.6; PTPN8

          View more View less

          UniProt ID: (Human) Q9Y2R2

          View more View less

          Entrez Gene ID: (Human) 26191

          View more View less

          Function(s)
          phosphoprotein phosphatase activity protein tyrosine phosphatase activity non-membrane spanning protein tyrosine phosphatase activity protein binding hydrolase activity phosphatase activity SH3 domain binding kinase binding ubiquitin protein ligase binding
          Process(es)
          immune system process lipid metabolic process autophagy negative regulation of autophagy positive regulation of gene expression negative regulation of gene expression T cell differentiation lipopolysaccharide-mediated signaling pathway positive regulation of type I interferon production response to lipopolysaccharide negative regulation of interleukin-6 production negative regulation of interleukin-8 production negative regulation of tumor necrosis factor production positive regulation of interferon-gamma production regulation of natural killer cell proliferation positive regulation of toll-like receptor 3 signaling pathway positive regulation of toll-like receptor 4 signaling pathway phosphoanandamide dephosphorylation negative regulation of JUN kinase activity regulation of innate immune response T cell receptor signaling pathway regulation of B cell receptor signaling pathway regulation of T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway negative regulation of T cell activation positive regulation of ERK1 and ERK2 cascade negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway cellular response to muramyl dipeptide positive regulation of NLRP3 inflammasome complex assembly regulation of NIK/NF-kappaB signaling positive regulation of protein K63-linked ubiquitination negative regulation of p38MAPK cascade
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline