Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CD148 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Abnova

          PTPRJ Polyclonal Antibody, MaxPab™

          View all (13) CD148 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support

          Cite PTPRJ Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PTPRJ Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PTPRJ Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTPRJ Antibody (H00005795-B01P) in WB

          Western Blot analysis of PTPRJ expression in transfected 293T cell line (H00005795-T01) by PTPRJ MaxPab polyclonal antibody.

          Lane 1: PTPRJ transfected lysate(147.1 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTPRJ Antibody in Western Blot (WB)
          PTPRJ Polyclonal Antibody, MaxPab™

          Product Details

          H00005795-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PTPRJ (AAI60125.1, 1 a.a. approximately 1337 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MKPAAREARL PPRSPGLRWA LPLLLLLLRL GQILCAGGTP SPIPDPSVAT VATGENGITQ ISSTAESFHK QNGTGTPQVE TNTSEDGESS GANDSLRTPE QGSNGTDGAS QKTPSSTGPS PVFDIKAVSI SPTNVILTWK SNDTAASEYK YVVKHKMENE KTITVVHQPW CNITGLRPAT SYVFSITPGI GNETWGDPRV IKVITEPIPV SDLRVALTGV RKAALSWSNG NGTASCRVLL ESIGSHEELT QDSRLQVNIS GLKPGVQYNI NPYLLQSNKT KGDPLGTEGG LDASNTERSR AGSPTAPVHD ESLVGPVDPS SGQQSRDTEV LLVGLEPGTR YNATVYSQAA NGTEGQPQAI EFRTNAIQVF DVTAVNISAT SLTLIWKVSD NESSSNYTYK IHVAGETDSS NLNVSEPRAV IPGLRSSTFY NITVCPVLGD IEGTPGFLQV HTPPVPVSDF RVTVVSTTEI GLAWSSHDAE SFQMHITQEG AGNSRVEITT NQSIIIGGLF PGTKYCFEIV PKGPNGTEGA SRTVCNRTVP SAVFDIHVVY VTTTEMWLDW KSPDGASEYV YHLVIESKHG SNHTSTYDKA ITLQGLIPGT LYNITISPEV DHVWGDPNST AQYTRPSNVS NIDVSTNTTA ATLSWQNFDD ASPTYSYCLL IEKAGNSSNA TQVVTDIGIT DATVTELIPG SSYTVEIFAQ VGDGIKSLEP GRKSFCTDPA SMASFDCEVV PKEPALVLKW TCPPGANAGF ELEVSSGAWN NATHLESCSS ENGTEYRTEV TYLNFSTSYN ISITTVSCGK MAAPTRNTCT TGITDPPPPD GSPNITSVSH NSVKVKFSGF EASHGPIKAY AVILTTGEAG HPSADVLKYT YEDFKKGASD TYVTYLIRTE EKGRSQSLSE VLKYEIDVGN ESTTLGYYNG KLEPLGSYRA CVAGFTNITF HPQNKGLIDG AESYVSFSRY SDAVSLPQDP GVICGAVFGC IFGALVIVTV GGFIFWRKKR KDAKNNEVSF SQIKPKKSKL IRVENFEAYF KKQQADSNCG FAEEYEDLKL VGISQPKYAA ELAENRGKNR YNNVLPYDIS RVKLSVQTHS TDDYINANYM PGYHSKKDFI ATQGPLPNTL KDFWRMVWEK NVYAIIMLTK CVEQGRTKCE EYWPSKQAQD YGDITVAMTS EIVLPEWTIR DFTVKNIQTS ESHPLRQFHF TSWPDHGVPD TTDLLINFRY LVRDYMKQSP PESPILVHCS AGVGRTGTFI AIDRLIYQIE NENTVDVYGI VYDLRMHRPL MVQTEDQYVF LNQCVLDIVR SQKDSKVDLI YQNTTAMTIY ENLAPVTTFG KTNGYIA

          Target Information

          CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline