Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PTPRN2 Antibodies

          Abnova

          PTPRN2 Polyclonal Antibody, MaxPab™

          View all (16) PTPRN2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PTPRN2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PTPRN2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PTPRN2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTPRN2 Antibody (H00005799-D01P) in WB

          Western Blot analysis of PTPRN2 expression in transfected 293T cell line by PTPRN2 MaxPab polyclonal antibody. Lane 1: PTPRN2 transfected lysate(108.10 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTPRN2 Antibody in Western Blot (WB)

          Product Details

          H00005799-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PTPRN2 (NP_570858.1, 1 a.a. approximately 986 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGPPLPLLLL LLLLLPPRVL PAAPSSVPRG RQLPGRLGCL LEEGLCGASE ACVNDGVFGR CQKVPAMDFY RYEVSPVALQ RLRVALQKLS GTGFTWQDDY TQYVMDQELA DLPKTYLRRP EASSPARPSK HSVGSERRYS REGGAALANA LRRHLPFLEA LSQAPASDVL ARTHTAQDRP PAEGDDRFSE SILTYVAHTS ALTYPPGPRT QLREDLLPRT LGQLQPDELS PKVDSGVDRH HLMAALSAYA AQRPPAPPGE GSLEPQYLLR APSRMPRPLL APAAPQKWPS PLGDSEDPSS TGDGARIHTL LKDLQRQPAE VRGLSGLELD GMAELMAGLM QGVDHGVARG SPGRAALGES GEQADGPKAT LRGDSFPDDG VQDDDDRLYQ EVHRLSATLG GLLQDHGSRL LPGALPFARP LDMERKKSEH PESSLSSEEE TAGVENVKSQ TYSKDLLGQQ PHSEPGAAAF GELQNQMPGP SKEEQSLPAG AQEALSDGLQ LEVQPSEEEA RGYIVTDREV LGPAVTFKVS ANVQNVTTED VEKATVDNKD KLEETSGLKI LQTGVGSKSK LKFLPPQAEQ EDSTKFIALT LVSLACILGV LLASGLIYCL RHSSQHRLKE KLSGLGGDPG ADATAAYQEL CRQRMATRPP DRPEGPHTSR ISSVSSQFSD GPIPSPSARS SASSWSEEPV QSNMDISTGH MILSYMEDHL KNKNRLEKEW EALCAYQAEP NSSFVAQREE NVPKNRSLAV LTYDHSRVLL KAENSHSHSD YINASPIMDH DPRNPAYIAT QGPLPATVAD FWQMVWESGC VVIVMLTPLA ENGVRQCYHY WPDEGSNLYH IYEVNLVSEH IWCEDFLVRS FYLKNLQTNE TRTVTQFHFL SWYDRGVPSS SRSLLDFRRK VNKCYRGRSC PIIVHCSDGA GRSGTYVLID MVLNKMAKGA KEIDIAATLE HLRDQRPGMV QTKEQFEFAL TAVAEEVNAI LKALPQ

          Target Information

          The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domain, and thus represents a receptor-type PTP. The catalytic domain of this PTP is most closely related to PTPRN/IA-2beta. This PTP and PTPRN are both found to be major autoantigens associated with insulin-dependent diabetes mellitus. Three alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EC 3.1.3.48; IAR; IAR/receptor-like protein-tyrosine phosphatase; Islet cell autoantigen-related protein; KIAA0387; Phogrin; protein tyrosine phosphatase homolog; protein tyrosine phosphatase receptor pi; protein tyrosine phosphatase, receptor type, N polypeptide 2; PTP-IA-2beta; transmembrane protein; R-PTP-N2; Receptor-type tyrosine-protein phosphatase N2; tyrosine phosphatase IA-2 beta

          View more View less

          Gene Aliases: IA-2beta; IAR; ICAAR; KIAA0387; PTPRN2; PTPRP; R-PTP-N2

          View more View less

          UniProt ID: (Human) Q92932

          View more View less

          Entrez Gene ID: (Human) 5799

          View more View less

          Function(s)
          phosphoprotein phosphatase activity protein tyrosine phosphatase activity transmembrane receptor protein tyrosine phosphatase activity hydrolase activity
          Process(es)
          protein dephosphorylation lipid metabolic process neurotransmitter secretion insulin secretion involved in cellular response to glucose stimulus regulation of secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline