Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Sodium Channel Pan Antibodies

          Alomone Labs, LTD

          Pan NaV Polyclonal Antibody

          View all (17) Sodium Channel Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Pan NaV Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Pan NaV Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Pan NaV Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Pan NaV Antibody (ASC-003-GP200UL) in WB

          Western blot analysis of rat brain (lanes 1 and 3) and mouse brain (lanes 2 and 4) lysates: - 1,2.Guinea pig Anti-Pan NaV Antibody (#ASC-003-GP), (1:800).3,4. Guinea pig Anti-Pan NaV Antibody , preincubated with Pan Nav Blocking Peptide (#BLP-SC003). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Pan NaV Antibody in Western Blot (WB)
          Pan NaV Polyclonal Antibody

          Product Details

          ASC-003-GP200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:800
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Guinea pig / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TEEQKKYYNAMKKLGSKK(C), corresponding to amino acid residues 1501-1518 of rat NaV1.1, Intracellular loop between domains III and IV
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Amount

          200 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          Voltage-gated sodium channels (NaV) are essential for the generation of action potentials and for cell excitability. NaV channels are activated in response to depolarization and selectively allow flow of Na+ ions. To date, nine NaV alpha subunits have been cloned and named NaV1.1-NaV1.9. The NaV channels are classified into two groups according to their sensitivity to tetrodotoxin (TTX): TTX-sensitive (NaV1.1, NaV1.2, NaV1.3, NaV1.4, NaV1.6 and NaV1.7) and TTX-resistant (NaV1.5, NaV1.8 and NaV1.9). Mammalian sodium channels are heterotrimers, composed of a central, pore-forming alpha subunit and two auxiliary beta subunits. The expression of the alpha subunit isoform is developmentally regulated and tissue specific. Sodium channels in the adult central nervous system and heart contain beta 1 through beta 4 subunits, whereas sodium channels in adult skeletal muscle have only the beta 1 subunit.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3-Feb; alternative product; ataxia 3; brain III voltage-gated sodium channel; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CTC-264K15.6; H1B; HBSC II; hH1; hH1c; voltage-gated sodium channel; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; mH1; Mu-1; Na+ channel; NaCh6; NaN; NaN alpha subunit; Nav1.4; Nav1.5/SCN5A.3; Neuroendocrine sodium channel; Peripheral nerve protein type 4; Peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; Peripheral sodium channel 1; PN1; PN3; PN4; PN5; RIIA sodium channel protein; SCN10A/PN3; scn5a-JEJ, HH1, hNav1.5, LQT3; SCN8A; Sensory neuron sodium channel; Sensory neuron sodium channel 2; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; SkM1; sodium channel 25; Sodium channel 6; sodium channel alpha-subunit; Sodium channel protein brain I subunit alpha; Sodium channel protein brain II subunit alpha; Sodium channel protein brain III subunit alpha; Sodium channel protein cardiac muscle subunit alpha; Sodium channel protein skeletal muscle subunit alpha; Sodium channel protein type 1 subunit alpha; Sodium channel protein type 10 subunit alpha; Sodium channel protein type 11 subunit alpha; Sodium channel protein type 2 subunit alpha; Sodium channel protein type 3 subunit alpha; Sodium channel protein type 4 subunit alpha; Sodium channel protein type 5 subunit alpha; Sodium channel protein type 8 subunit alpha; Sodium channel protein type 9 subunit alpha; Sodium channel protein type I subunit alpha; Sodium channel protein type II subunit alpha; Sodium channel protein type III subunit alpha; Sodium channel protein type IV subunit alpha; Sodium channel protein type IX subunit alpha; Sodium channel protein type V subunit alpha; Sodium channel protein type VIII subunit alpha; Sodium channel protein type X subunit alpha; Sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain II subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel type X alpha polypeptide; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type 4 alpha polypeptide; sodium channel voltage-gated type I alpha polypeptide; sodium channel voltage-gated type II alpha polypeptide; Sodium channel voltage-gated type IV alpha polypeptide; sodium channel voltage-gated type IX alpha; sodium channel voltage-gated type V alpha polypeptide; sodium channel voltage-gated type VIII alpha; sodium channel voltage-gated type VIII alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage-gated, type 1, alpha polypeptide; sodium channel, voltage-gated, type 10, alpha polypeptide; sodium channel, voltage-gated, type 11, alpha polypeptide; sodium channel, voltage-gated, type 2, alpha 1 polypeptide; sodium channel, voltage-gated, type 2, alpha 1 subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type 4, alpha polypeptide; sodium channel, voltage-gated, type 4, alpha subunit; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type 8, alpha polypeptide; sodium channel, voltage-gated, type 8, alpha subunit; sodium channel, voltage-gated, type 9, alpha polypeptide; sodium channel, voltage-gated, type I, alpha polypeptide; sodium channel, voltage-gated, type I, alpha subunit; sodium channel, voltage-gated, type II, alpha 1; sodium channel, voltage-gated, type II, alpha 1 polypeptide; sodium channel, voltage-gated, type II, alpha 2 polypeptide; sodium channel, voltage-gated, type II, alpha polypeptide 1; sodium channel, voltage-gated, type II, alpha subunit; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha subunit; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; sodium channel, voltage-gated, type VIII, alpha polypeptide; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium channel, voltage-gated, type XI, alpha; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium channel, voltage-gated, type11, alpha polypeptide; SP-19; unnamed protein product; voltage gated sodium channel; voltage-gated sodium channel alpha subunit Nav1.7; voltage-gated sodium channel Nav1.5c; voltage-gated sodium channel NAV1.9b; voltage-gated sodium channel subtype II; Voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.1; Voltage-gated sodium channel subunit alpha Nav1.2; Voltage-gated sodium channel subunit alpha Nav1.3; Voltage-gated sodium channel subunit alpha Nav1.4; Voltage-gated sodium channel subunit alpha Nav1.5; Voltage-gated sodium channel subunit alpha Nav1.6; Voltage-gated sodium channel subunit alpha Nav1.7; Voltage-gated sodium channel subunit alpha Nav1.8; Voltage-gated sodium channel subunit alpha Nav1.9; voltage-gated sodium channel type II alpha subunit; voltage-gated sodium channel type VIII alpha protein; voltage-gated sodium channel; alpha subunit domain 4, S5-S6

          View more View less

          Gene Aliases: 6430408L10; A230052E19Rik; AI853486; B230332M13; BFIC3; BFIS3; BFIS5; BFNIS; C630029C19Rik; CDCD2; CERIII; CIAT; CMD1E; CMPD2; CMS16; CMYO22A; CMYP22A; DEE11; DEE13; DEE6; DEE62; DEE6A; DEE6B; dmu; DRVT; EA9; EIEE11; EIEE13; EIEE6; EIEE62; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FFEVF4; FHM3; GEFSP2; GEFSP7; Gm1000; HB1; HB2; HBA; HBBD; HBSCI; HBSCII; HH1; HOKPP2; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; Kiaa4197; LQT3; MED; mH1; mH2; microI; mKIAA4197; mnd-2; mnd2; MYOCL2; Na(v)1.2; Na(V)1.4; Na(V)1.8; NAC1; NAC1A; NAC2; NAC3; NaCh6; NachII; NaN; NaT; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.5c; Nav1.6; Nav1.7; Nav1.8; NAV1.9; Nbna1; NCHVS; NE-NA; NENA; nmf2; nmf335; nmf58; NSS2; nur14; PFHB1; PN1; PN3; PN4; PN5; RII/RIIA; RNSCPIIR; SCAL; SCIII; SCN; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; SCN9A; ScpII; seal; SFNP; SkM1; SkM2; SMEI; SNS; SNS-2; SNS2; SSS1; VF1

          View more View less

          UniProt ID: (Human) Q9Y5Y9, (Human) Q9UI33, (Human) P35498, (Human) Q99250, (Human) Q9NY46, (Human) P35499, (Human) Q14524, (Human) Q9UQD0, (Human) Q15858, (Rat) Q62968, (Mouse) Q6QIY3, (Rat) O88457, (Mouse) Q9R053, (Rat) P04774, (Mouse) A2APX8, (Rat) P04775, (Mouse) B1AWN6, (Mouse) A2ASI5, (Rat) P08104, (Mouse) Q9ER60, (Rat) P15390, (Rat) P15389, (Mouse) Q9JJV9, (Mouse) Q9WTU3, (Rat) O88420, (Mouse) Q62205, (Rat) O08562

          View more View less

          Entrez Gene ID: (Human) 6336, (Human) 11280, (Human) 6323, (Human) 6326, (Human) 6328, (Human) 6329, (Human) 6331, (Human) 6334, (Human) 6335, (Rat) 29571, (Mouse) 20264, (Rat) 29701, (Mouse) 24046, (Rat) 81574, (Mouse) 20265, (Rat) 24766, (Mouse) 110876, (Mouse) 20269, (Rat) 497770, (Mouse) 110880, (Rat) 25722, (Rat) 25665, (Mouse) 20271, (Mouse) 20273, (Rat) 29710, (Mouse) 20274, (Rat) 78956

          View more View less

          Function(s)
          ion channel activity voltage-gated sodium channel activity cation channel activity sodium channel activity ion channel binding voltage-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential protein binding calmodulin binding fibroblast growth factor binding enzyme binding protein kinase binding protein domain specific binding ankyrin binding ubiquitin protein ligase binding nitric-oxide synthase binding voltage-gated sodium channel activity involved in cardiac muscle cell action potential voltage-gated sodium channel activity involved in AV node cell action potential voltage-gated sodium channel activity involved in bundle of His cell action potential voltage-gated sodium channel activity involved in Purkinje myocyte action potential voltage-gated sodium channel activity involved in SA node cell action potential scaffold protein binding sodium ion binding voltage-gated ion channel activity leucine zipper domain binding nucleotide binding ATP binding voltage-gated ion channel
          Process(es)
          regulation of heart rate ion transport sodium ion transport sensory perception ion transmembrane transport regulation of ion transmembrane transport sodium ion transmembrane transport odontogenesis of dentin-containing tooth transmembrane transport regulation of cardiac muscle contraction regulation of atrial cardiac muscle cell membrane depolarization cardiac muscle cell action potential involved in contraction membrane depolarization during action potential AV node cell action potential bundle of His cell action potential regulation of presynaptic membrane potential action potential thigmotaxis acute inflammatory response chronic inflammatory response inflammatory response G-protein coupled receptor signaling pathway axonogenesis circadian rhythm chemosensory behavior response to heat response to cold response to xenobiotic stimulus response to toxic substance response to high light intensity response to auditory stimulus transmission of nerve impulse neuronal action potential sensory perception of pain response to prostaglandin E thermosensory behavior regulation of membrane potential mast cell degranulation response to pain behavioral response to pain cell motility detection of temperature stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception establishment of localization in cell reflex micturition skeletal muscle organ development artery development behavioral response to acetic acid induced pain behavioral response to formalin induced pain cellular response to cold calcium ion transmembrane transport response to nitric oxide action potential initiation cAMP/PKA signal transduction sensory perception of itch calcitonin gene-related peptide receptor signaling pathway small intestine smooth muscle contraction adult walking behavior determination of adult lifespan neuronal action potential propagation nerve development neuromuscular process controlling posture nervous system development memory intrinsic apoptotic signaling pathway in response to osmotic stress myelination neuron apoptotic process cellular response to hypoxia muscle contraction regulation of skeletal muscle contraction by action potential cardiac conduction system development cardiac ventricle development brainstem development positive regulation of sodium ion transport response to denervation involved in regulation of muscle adaptation telencephalon development cerebellum development positive regulation of action potential positive regulation of epithelial cell proliferation membrane depolarization cardiac muscle contraction regulation of ventricular cardiac muscle cell membrane repolarization regulation of atrial cardiac muscle cell membrane repolarization regulation of ventricular cardiac muscle cell membrane depolarization cellular response to calcium ion regulation of cardiac muscle cell contraction ventricular cardiac muscle cell action potential membrane depolarization during cardiac muscle cell action potential atrial cardiac muscle cell action potential SA node cell action potential membrane depolarization during AV node cell action potential membrane depolarization during SA node cell action potential membrane depolarization during Purkinje myocyte cell action potential membrane depolarization during bundle of His cell action potential AV node cell to bundle of His cell communication regulation of heart rate by cardiac conduction membrane depolarization during atrial cardiac muscle cell action potential regulation of sodium ion transmembrane transport peripheral nervous system development post-embryonic development action potential propagation cardiac conduction bundle of His cell to Purkinje myocyte communication transport regulation of sensory perception of pain positive regulation of defense response to virus by host response to pyrethroid cellular response to antibiotic response to wounding cation transport potassium ion transport choline transport positive regulation of heart rate response to organic cyclic compound SA node cell to atrial cardiac muscle cell communication muscle organ development sensory perception of sound locomotory behavior adult locomotory behavior neuromuscular process rhythmic excitation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline