Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Pan Ras Antibodies

          Invitrogen

          Pan Ras Polyclonal Antibody

          View all (11) Pan Ras antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Pan Ras Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Pan Ras Antibody (PA5-104464) in ICC/IF

          Immunofluorescent analysis of Pan Ras in HepG2. Samples were fixed with paraformaldehyde, permeabilized with 0.1% Triton X-100, blocked with 10% serum (45 min at 25°C), incubated with mouse anti-beta tubulin and Pan Ras polyclonal antibody (Product # PA5-104464) using a dilution of 1:200 (1 hr, 37°C), and followed by goat anti-rabbit IgG Alexa Fluor 594 (red) and goat anti-mouse IgG Alexa Fluor 488 (green). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Pan Ras Antibody in Immunocytochemistry (ICC/IF)
          Pan Ras Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Ras Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Pan Ras Antibody in Western Blot (WB)
          Pan Ras Antibody in Western Blot (WB)
          Pan Ras Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Pan Ras Polyclonal Antibody

          Product Details

          PA5-104464

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:300
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human NRAS(Accession P01111), corresponding to amino acid residues V7-D57.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2853765

          Product Specific Information

          Antibody detects endogenous levels of total RASH/RASK/RASN.

          Target Information

          This gene belongs to the Ras oncogene family, whose members are related to the transforming genes of mammalian sarcoma retroviruses. The products encoded by these genes function in signal transduction pathways. These proteins can bind GTP and GDP, and they have intrinsic GTPase activity. This protein undergoes a continuous cycle of de- and re-palmitoylation, which regulates its rapid exchange between the plasma membrane and the Golgi apparatus. Mutations in this gene cause Costello syndrome, a disease characterized by increased growth at the prenatal stage, growth deficiency at the postnatal stage, predisposition to tumor formation, mental retardation, skin and musculoskeletal abnormalities, distinctive facial appearance and cardiovascular abnormalities. Defects in this gene are implicated in a variety of cancers, including bladder cancer, follicular thyroid cancer, and oral squamous cell carcinoma. Multiple transcript variants, which encode different isoforms, have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: c-Ha-ras p21 protein; c-Ha-ras transgene; c-has/bas p21 protein; c-K-ras; c-K-ras2 protein; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-ras-Ki-2 protein; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; cellular transforming proto-oncogene; GTP- and GDP-binding peptide B; GTPase Hras1; H-ras 1 protein; H-Ras-1; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; HRAS; K RAS; K-Ras 2; K-ras p21 protein; ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); Kirsten rat sarcoma viral proto-oncogene; KRAS-ENOPH fusion; Kras-Enoph1 fusion protein; N-ras protein part 4; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; Nras; oncogene KRAS2; p19 H-RasIDX protein; p21 protein; p21ras; PR310 c-K-ras oncogene; PR371 c-K-ras oncogene; proto-oncogene GTPase; Ras family small GTP binding protein H-Ras; RASK; RASN; small GTP binding protein; transformation gene: oncogene HAMSV; transforming protein N-Ras; transforming protein P21; unnamed protein product; v-Ha-ras Harvey rat sarcoma viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; v-ras neuroblastoma RAS viral oncogene homolog

          View more View less

          Gene Aliases: 'C-K-RAS; ALPS4; C-BAS/HAS; C-H-RAS; c-Ha-ras; C-HA-RAS1; C-K-RAS; c-Ki-ras; c-Ki-ras2; c-rasHa; CFC2; CMNS; CTLO; H-ras; H-RASIDX; Ha-ras; HAMSV; Harvey-ras; Hras-1; HRAS1; K-Ras; K-Ras 2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; Kras-2; KRAS1; KRAS2; N-ras; NCMS; NRAS1; NS; NS3; NS6; OES; p21; p21B; p21ras; RALD; RALD1; RALD2; ras; RASH1; RASK2

          View more View less

          UniProt ID: (Human) P01112, (Mouse) Q61411, (Rat) P20171, (Human) P01116, (Mouse) P32883, (Rat) P08644, (Human) P01111, (Rat) Q04970

          View more View less

          Entrez Gene ID: (Human) 3265, (Mouse) 15461, (Rat) 293621, (Human) 3845, (Mouse) 16653, (Rat) 24525, (Human) 4893, (Mouse) 18176, (Rat) 24605

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding GDP binding protein anchor phospholipase C activator activity macromolecular complex binding GMP binding LRR domain binding identical protein binding
          Process(es)
          signal transduction I-kappaB kinase/NF-kappaB signaling Ras protein signal transduction positive regulation of cell proliferation visual learning gene expression positive regulation of gene expression cytokine-mediated signaling pathway forebrain astrocyte development actin cytoskeleton organization regulation of synaptic transmission, GABAergic positive regulation of Rac protein signal transduction positive regulation of MAP kinase activity negative regulation of neuron apoptotic process negative regulation of cell differentiation insulin-like growth factor receptor signaling pathway regulation of long-term neuronal synaptic plasticity homeostasis of number of cells within a tissue positive regulation of nitric-oxide synthase activity striated muscle cell differentiation epithelial tube branching involved in lung morphogenesis positive regulation of cellular senescence liver development female pregnancy response to gravity response to isolation stress response to glucocorticoid response to mineralocorticoid myoblast proliferation cardiac muscle cell proliferation MAPK cascade regulation of transcription from RNA polymerase II promoter chemotaxis cell surface receptor signaling pathway negative regulation of cell proliferation insulin receptor signaling pathway organ morphogenesis negative regulation of gene expression Schwann cell development positive regulation of cell migration regulation of actin cytoskeleton organization regulation of cell proliferation myelination positive regulation of MAPK cascade positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of Ras protein signal transduction positive regulation of epithelial cell proliferation regulation of cell cycle adipose tissue development positive regulation of ERK1 and ERK2 cascade cellular response to gamma radiation positive regulation of wound healing positive regulation of protein targeting to membrane cellular senescence regulation of neurotransmitter receptor localization to postsynaptic specialization membrane positive regulation of ruffle assembly positive regulation of miRNA metabolic process regulation of protein phosphorylation positive regulation of endothelial cell proliferation regulation of gene expression positive regulation of interferon-gamma production defense response to protozoan positive regulation of angiogenesis positive regulation of fibroblast proliferation negative regulation of skeletal muscle tissue development T cell receptor signaling pathway myoblast differentiation positive regulation of TOR signaling negative regulation of programmed cell death endocytosis small GTPase mediated signal transduction ERBB2-ERBB3 signaling pathway T-helper 1 type immune response oncogene-induced cell senescence intrinsic apoptotic signaling pathway positive regulation of DNA replication
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-hhljg:80/100.66.77.101:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0