Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Phospho-EphB1/EphB3/EphB4 (Tyr778, Tyr792, Tyr774) Polyclonal Antibody detects endogenous levels of EphB1/EphB3/EphB4 only when phosphorylated at Tyr778, Tyr792, Tyr774.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Developmental kinase 2; Developmental kinase 5; EK2; EK6; ELK; ELK-related protein tyrosine kinase; Embryonic kinase 2; Eph receptor B2 (ELK-related protein tyrosine kinase); EPH receptor B4; EPH tyrosine kinase 2; EPH-like kinase 2; EPH-like kinase 6; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; ephrin receptor EphB4; Ephrin type-B receptor 1; Ephrin type-B receptor 3; Ephrin type-B receptor 4; Hepatoma transmembrane kinase; human embryo kinase 2; mDK-2; mDK-5; NET; Neuronally-expressed EPH-related tyrosine kinase; soluble EPHB1 variant 1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; Tyrosine kinase MYK-1; Tyrosine-protein kinase receptor EPH-2; tyrosine-protein kinase receptor HTK; Tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO11; Tyrosine-protein kinase TYRO6
Gene Aliases: 9330129L11; AI042935; AW456895; AW488255; C130099E04Rik; Cek10; Cek6; EK2; ELK; Elkh; ENSMUSG00000074119; EPHB1; Ephb2; EPHB3; EPHB4; EPHT2; Epth2; Erk; ETK2; HEK2; HEK6; HTK; MDK2; MDK5; MYK1; NET; Sek4; TYRO11; TYRO6
UniProt ID: (Human) P54762, (Human) P54753, (Human) P54760, (Rat) P09759, (Mouse) Q8CBF3, (Mouse) P54754, (Mouse) P54761
Entrez Gene ID: (Human) 2047, (Human) 2049, (Human) 2050, (Rat) 24338, (Mouse) 270190, (Mouse) 13845, (Rat) 287989, (Rat) 686310, (Mouse) 13846
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support