Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Phospho-NDRG2 (Thr348) Polyclonal Antibody detects endogenous levels of NDRG2 only when phosphorylated at Thr348.
NDRG2's gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antidepressant-related protein ADRG123; cytoplasmic protein Ndr1; DKFZp781G1938; FLJ25522; N-myc downstream regulated 2; N-myc downstream regulated gene 2; N-myc downstream regulator 2; N-myc downstream-regulated gene 2 protein; NDR1-related protein NDR2; NDRG1-related protein; NDRG2var; Protein Ndr2; Protein NDRG2; Protein Syld709613; syld709613 protein
Gene Aliases: AI182517; AU040374; KIAA1248; Ndr2; NDRG2; SYLD
UniProt ID: (Human) Q9UN36, (Mouse) Q9QYG0, (Rat) Q8VBU2
Entrez Gene ID: (Human) 57447, (Mouse) 29811, (Rat) 171114
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support