Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:2,500-1:10,000 | - |
Immunocytochemistry (ICC/IF) |
1:1500-1:50000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
A synthetic peptide corresponding to the center region of the Human PRKRA |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
Lot-Specific |
Purification |
Protein A, Antigen affinity chromatography |
Storage buffer |
PBS |
Contains |
no preservative |
Storage conditions |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
RRID |
AB_2788470 |
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human PACT/PRKRA.
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: interferon-inducible double stranded RNA-dependent protein kinase activator A; Interferon-inducible double-stranded RNA-dependent protein kinase activator A; PKR-associated protein X; PKR-associating protein X; Protein activator of the interferon-induced protein kinase; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; Protein kinase, interferon-inducible double-stranded RNA-dependent activator
Gene Aliases: DYT16; HSD-14; HSD14; PACT; PRKRA; RAX
UniProt ID: (Human) O75569
Entrez Gene ID: (Human) 8575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support